Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 56460190-56460317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DST
Entrez Gene ID
667
Gene Name
dystonin
Gene Aliases
BP240, BPA, BPAG1, CATX-15, CATX15, D6S1101, DMH, DT, EBSB2, HSAN6, MACF2
UniGene
Hs.604915
Gene Chromosome Location
Chr.6: 56457987-56954671 on Build GRCh38
Transcripts
NM_001144769.2 NM_001144770.1 NM_015548.4 NM_183380.3 XM_005249310.3 XM_005249315.3 XM_005249316.2 XM_005249318.2 XM_005249319.2 XM_005249320.4 XM_005249322.4 XM_005249323.3 XM_005249324.2 XM_011514824.2 XM_011514825.2 XM_011514826.2 XM_017011205.1 XM_017011206.1 XM_017011207.1 XM_017011208.1 XM_017011209.1 XM_017011210.1 XM_017011211.1 XM_017011212.1 XM_017011213.1 XM_017011214.1 XM_017011215.1 XM_017011216.1 XM_017011217.1 XM_017011218.1 XM_017011219.1 XM_017011220.1 XM_017011221.1 XM_017011222.1 XM_017011223.1 XM_017011224.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
421261 c.22283T>A p.L7428H Chr.6: 56460223-56460223 on Build GRCh38
5530224 c.15189C>T p.F5063F Chr.6: 56460195-56460195 on Build GRCh38
421260 c.15161T>A p.L5054H Chr.6: 56460223-56460223 on Build GRCh38
395344 c.15144A>C p.Q5048H Chr.6: 56460240-56460240 on Build GRCh38
4586851 c.22275G>A p.K7425K Chr.6: 56460231-56460231 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770294227 Chr.6: 56460247-56460247 on Build GRCh38
rs372639720 Chr.6: 56460226-56460226 on Build GRCh38
rs375362610 Chr.6: 56460249-56460249 on Build GRCh38
rs750247142 Chr.6: 56460258-56460259 on Build GRCh38
rs377080714 Chr.6: 56460231-56460231 on Build GRCh38
rs771734453 Chr.6: 56460267-56460267 on Build GRCh38
rs760404859 Chr.6: 56460276-56460276 on Build GRCh38
rs780195917 Chr.6: 56460210-56460210 on Build GRCh38
rs368992091 Chr.6: 56460216-56460216 on Build GRCh38
rs759079609 Chr.6: 56460292-56460292 on Build GRCh38
rs753412060 Chr.6: 56460289-56460289 on Build GRCh38
rs532258725 Chr.6: 56460250-56460250 on Build GRCh38
rs775047126 Chr.6: 56460275-56460275 on Build GRCh38
rs370496665 Chr.6: 56460246-56460246 on Build GRCh38
rs763646862 Chr.6: 56460278-56460278 on Build GRCh38

View Full Product Details