Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3153460-3153612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRBN
Entrez Gene ID
51185
Gene Name
cereblon
Gene Aliases
MRT2, MRT2A
UniGene
Hs.18925
Gene Chromosome Location
Chr.3: 3148490-3179717 on Build GRCh38
Transcripts
NM_001173482.1 NM_016302.3 XM_005265202.3 XM_011533791.2 XM_011533793.2 XM_011533794.2

SNP Detail

SNP ID Chromosome Location
rs569745851 Chr.3: 3153519-3153519 on Build GRCh38
rs905902110 Chr.3: 3153576-3153576 on Build GRCh38
rs763155197 Chr.3: 3153530-3153530 on Build GRCh38
rs865869655 Chr.3: 3153585-3153585 on Build GRCh38
rs760603312 Chr.3: 3153501-3153501 on Build GRCh38
rs773456257 Chr.3: 3153525-3153525 on Build GRCh38
rs1009525432 Chr.3: 3153554-3153554 on Build GRCh38
rs776654473 Chr.3: 3153516-3153516 on Build GRCh38
rs376683996 Chr.3: 3153520-3153520 on Build GRCh38
rs370167783 Chr.3: 3153491-3153491 on Build GRCh38
rs892379739 Chr.3: 3153541-3153541 on Build GRCh38
rs1048867153 Chr.3: 3153561-3153561 on Build GRCh38
rs931420102 Chr.3: 3153490-3153490 on Build GRCh38
rs538707484 Chr.3: 3153569-3153569 on Build GRCh38

View Full Product Details