Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 47375108-47375275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMPK1
Entrez Gene ID
51727
Gene Name
cytidine/uridine monophosphate kinase 1
Gene Aliases
CK, CMK, CMPK, UMK, UMP-CMPK, UMPK
UniGene
Hs.731647
Gene Chromosome Location
Chr.1: 47333797-47378839 on Build GRCh38
Transcripts
NM_001136140.1 NM_016308.2 NR_046394.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
398125 c.572C>A p.T191K Chr.1: 47375220-47375220 on Build GRCh38
4008644 c.555G>A p.Q185Q Chr.1: 47375203-47375203 on Build GRCh38
1343213 c.549-2A>G p.? Chr.1: 47375195-47375195 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778487021 Chr.1: 47375158-47375165 on Build GRCh38
rs768266149 Chr.1: 47375161-47375161 on Build GRCh38
rs935377143 Chr.1: 47375185-47375187 on Build GRCh38
rs373906378 Chr.1: 47375176-47375176 on Build GRCh38
rs915362521 Chr.1: 47375235-47375235 on Build GRCh38
rs768774978 Chr.1: 47375171-47375171 on Build GRCh38
rs72553950 Chr.1: 47375138-47375138 on Build GRCh38
rs781755082 Chr.1: 47375158-47375158 on Build GRCh38
rs1041301166 Chr.1: 47375142-47375142 on Build GRCh38
rs749505722 Chr.1: 47375151-47375151 on Build GRCh38
rs562718888 Chr.1: 47375173-47375173 on Build GRCh38
rs748765179 Chr.1: 47375149-47375152 on Build GRCh38
rs1052529462 Chr.1: 47375188-47375188 on Build GRCh38
rs756932122 Chr.1: 47375156-47375159 on Build GRCh38
rs771468808 Chr.1: 47375147-47375147 on Build GRCh38
rs370461272 Chr.1: 47375146-47375146 on Build GRCh38
rs772791328 Chr.1: 47375210-47375210 on Build GRCh38
rs746444235 Chr.1: 47375160-47375160 on Build GRCh38
rs771705851 Chr.1: 47375191-47375191 on Build GRCh38

View Full Product Details