Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 163439152-163439403 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNG1
Entrez Gene ID
900
Gene Name
cyclin G1
Gene Aliases
CCNG
UniGene
Hs.79101
Gene Chromosome Location
Chr.5: 163437567-163445016 on Build GRCh38
Transcripts
NM_004060.3 NM_199246.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634120 c.28T>A p.S10T Chr.5: 163439284-163439284 on Build GRCh38
4404167 c.64G>A p.E22K Chr.5: 163439320-163439320 on Build GRCh38
736894 c.46C>A p.Q16K Chr.5: 163439302-163439302 on Build GRCh38
7195880 c.103A>T p.R35* Chr.5: 163439359-163439359 on Build GRCh38
1661043 c.90G>T p.K30N Chr.5: 163439346-163439346 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747141558 Chr.5: 163439271-163439271 on Build GRCh38
rs987385048 Chr.5: 163439311-163439311 on Build GRCh38
rs187330500 Chr.5: 163439186-163439186 on Build GRCh38
rs763341307 Chr.5: 163439315-163439315 on Build GRCh38
rs752347567 Chr.5: 163439236-163439236 on Build GRCh38
rs770005853 Chr.5: 163439302-163439302 on Build GRCh38
rs751113628 Chr.5: 163439225-163439225 on Build GRCh38
rs568802529 Chr.5: 163439191-163439191 on Build GRCh38
rs757258231 Chr.5: 163439265-163439265 on Build GRCh38
rs139729782 Chr.5: 163439310-163439310 on Build GRCh38
rs767222349 Chr.5: 163439235-163439235 on Build GRCh38
rs778976832 Chr.5: 163439370-163439370 on Build GRCh38
rs749138963 Chr.5: 163439249-163439249 on Build GRCh38
rs751598017 Chr.5: 163439248-163439249 on Build GRCh38
rs957780722 Chr.5: 163439205-163439205 on Build GRCh38
rs777642745 Chr.5: 163439238-163439238 on Build GRCh38
rs373860207 Chr.5: 163439277-163439277 on Build GRCh38
rs763783986 Chr.5: 163439240-163439240 on Build GRCh38
rs375674960 Chr.5: 163439322-163439322 on Build GRCh38
rs767738364 Chr.5: 163439368-163439368 on Build GRCh38
rs373954399 Chr.5: 163439325-163439325 on Build GRCh38
rs763543170 Chr.5: 163439213-163439213 on Build GRCh38
rs750393158 Chr.5: 163439379-163439379 on Build GRCh38
rs755843330 Chr.5: 163439237-163439237 on Build GRCh38
rs201194575 Chr.5: 163439347-163439347 on Build GRCh38
rs540263138 Chr.5: 163439185-163439185 on Build GRCh38
rs754605512 Chr.5: 163439233-163439233 on Build GRCh38
rs748394523 Chr.5: 163439288-163439288 on Build GRCh38
rs368403737 Chr.5: 163439360-163439360 on Build GRCh38
rs757231054 Chr.5: 163439307-163439316 on Build GRCh38
rs781444433 Chr.5: 163439380-163439380 on Build GRCh38
rs1026597944 Chr.5: 163439353-163439353 on Build GRCh38
rs767024264 Chr.5: 163439323-163439323 on Build GRCh38
rs145299150 Chr.5: 163439345-163439345 on Build GRCh38
rs776928202 Chr.5: 163439286-163439286 on Build GRCh38
rs531008144 Chr.5: 163439189-163439189 on Build GRCh38
rs766962044 Chr.5: 163439316-163439316 on Build GRCh38
rs760406711 Chr.5: 163439334-163439334 on Build GRCh38
rs1050012999 Chr.5: 163439247-163439247 on Build GRCh38
rs765771131 Chr.5: 163439219-163439219 on Build GRCh38
rs550507893 Chr.5: 163439179-163439179 on Build GRCh38
rs779973196 Chr.5: 163439269-163439269 on Build GRCh38

View Full Product Details