Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13918703-13918976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CC2D1A
Entrez Gene ID
54862
Gene Name
coiled-coil and C2 domain containing 1A
Gene Aliases
FREUD-1, Freud-1/Aki1, MRT3
UniGene
Hs.269592
Gene Chromosome Location
Chr.19: 13906143-13930880 on Build GRCh38
Transcripts
NM_017721.4 XM_005259972.2 XM_005259973.2 XM_005259974.2 XM_005259975.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6383229 c.1012A>T p.T338S Chr.19: 13918811-13918811 on Build GRCh38
6083694 c.980C>T p.S327L Chr.19: 13918779-13918779 on Build GRCh38
438701 c.1081G>C p.V361L Chr.19: 13918974-13918974 on Build GRCh38
4923348 c.1021G>T p.V341L Chr.19: 13918914-13918914 on Build GRCh38
3822084 c.1064G>A p.R355Q Chr.19: 13918957-13918957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs968306509 Chr.19: 13918878-13918878 on Build GRCh38
rs547816872 Chr.19: 13918790-13918790 on Build GRCh38
rs45509395 Chr.19: 13918726-13918726 on Build GRCh38
rs369829647 Chr.19: 13918914-13918914 on Build GRCh38
rs768483916 Chr.19: 13918946-13918946 on Build GRCh38
rs1056103923 Chr.19: 13918905-13918905 on Build GRCh38
rs770737062 Chr.19: 13918807-13918807 on Build GRCh38
rs1038583292 Chr.19: 13918768-13918768 on Build GRCh38
rs989389467 Chr.19: 13918772-13918780 on Build GRCh38
rs765282375 Chr.19: 13918758-13918758 on Build GRCh38
rs986781288 Chr.19: 13918767-13918767 on Build GRCh38
rs777294523 Chr.19: 13918895-13918895 on Build GRCh38
rs756554308 Chr.19: 13918800-13918800 on Build GRCh38
rs948919266 Chr.19: 13918827-13918827 on Build GRCh38
rs745631694 Chr.19: 13918927-13918927 on Build GRCh38
rs754413580 Chr.19: 13918889-13918889 on Build GRCh38
rs373278035 Chr.19: 13918938-13918938 on Build GRCh38
rs780393924 Chr.19: 13918861-13918861 on Build GRCh38
rs776586922 Chr.19: 13918736-13918736 on Build GRCh38
rs759464814 Chr.19: 13918816-13918816 on Build GRCh38
rs766421763 Chr.19: 13918732-13918733 on Build GRCh38
rs201374643 Chr.19: 13918796-13918796 on Build GRCh38
rs761958009 Chr.19: 13918832-13918832 on Build GRCh38
rs371690822 Chr.19: 13918857-13918857 on Build GRCh38
rs775171615 Chr.19: 13918942-13918942 on Build GRCh38
rs771652291 Chr.19: 13918803-13918803 on Build GRCh38
rs558746804 Chr.19: 13918920-13918920 on Build GRCh38
rs762923544 Chr.19: 13918826-13918826 on Build GRCh38
rs868611556 Chr.19: 13918926-13918926 on Build GRCh38
rs753784103 Chr.19: 13918885-13918886 on Build GRCh38
rs900587111 Chr.19: 13918761-13918761 on Build GRCh38
rs576388238 Chr.19: 13918945-13918945 on Build GRCh38
rs772713840 Chr.19: 13918804-13918804 on Build GRCh38
rs928862667 Chr.19: 13918892-13918892 on Build GRCh38
rs1022007374 Chr.19: 13918846-13918846 on Build GRCh38
rs746607724 Chr.19: 13918805-13918805 on Build GRCh38
rs372403900 Chr.19: 13918817-13918817 on Build GRCh38
rs548617953 Chr.19: 13918828-13918828 on Build GRCh38
rs200121704 Chr.19: 13918779-13918779 on Build GRCh38
rs781498477 Chr.19: 13918791-13918791 on Build GRCh38
rs61504820 Chr.19: 13918845-13918845 on Build GRCh38
rs781060128 Chr.19: 13918924-13918924 on Build GRCh38
rs202106696 Chr.19: 13918789-13918789 on Build GRCh38
rs756537978 Chr.19: 13918855-13918855 on Build GRCh38
rs537546040 Chr.19: 13918904-13918904 on Build GRCh38
rs759503584 Chr.19: 13918743-13918743 on Build GRCh38
rs758574171 Chr.19: 13918762-13918762 on Build GRCh38
rs11883041 Chr.19: 13918814-13918814 on Build GRCh38
rs372082075 Chr.19: 13918801-13918801 on Build GRCh38
rs750771780 Chr.19: 13918848-13918848 on Build GRCh38
rs751622351 Chr.19: 13918831-13918831 on Build GRCh38
rs887935645 Chr.19: 13918928-13918928 on Build GRCh38
rs866681046 Chr.19: 13918742-13918742 on Build GRCh38

View Full Product Details