Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 112233155-112233424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBLN7
Entrez Gene ID
129804
Gene Name
fibulin 7
Gene Aliases
TM14
UniGene
Hs.437696
Gene Chromosome Location
Chr.2: 112138385-112254888 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Gene Symbol
ZC3H8
Entrez Gene ID
84524
Gene Name
zinc finger CCCH-type containing 8
Gene Aliases
Fliz1, ZC3HDC8
UniGene
Hs.418416
Gene Chromosome Location
Chr.2: 112215035-112255087 on Build GRCh38
Transcripts
XM_017003317.1 NM_032494.2 XM_011511998.1 XM_017005107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3039417 c.687T>A p.Y229* Chr.2: 112233306-112233306 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs892874340 Chr.2: 112233385-112233385 on Build GRCh38
rs373897826 Chr.2: 112233246-112233246 on Build GRCh38
rs774751595 Chr.2: 112233361-112233361 on Build GRCh38
rs548005710 Chr.2: 112233384-112233384 on Build GRCh38
rs533360084 Chr.2: 112233207-112233207 on Build GRCh38
rs546630580 Chr.2: 112233224-112233224 on Build GRCh38
rs759954027 Chr.2: 112233363-112233363 on Build GRCh38
rs138032670 Chr.2: 112233341-112233341 on Build GRCh38
rs763254522 Chr.2: 112233230-112233231 on Build GRCh38
rs200470552 Chr.2: 112233335-112233335 on Build GRCh38
rs751854093 Chr.2: 112233204-112233204 on Build GRCh38
rs374265626 Chr.2: 112233374-112233374 on Build GRCh38
rs368041966 Chr.2: 112233248-112233248 on Build GRCh38
rs750192876 Chr.2: 112233226-112233226 on Build GRCh38
rs746930630 Chr.2: 112233242-112233242 on Build GRCh38
rs35782954 Chr.2: 112233273-112233273 on Build GRCh38
rs766588146 Chr.2: 112233232-112233233 on Build GRCh38
rs368474720 Chr.2: 112233375-112233375 on Build GRCh38
rs758298132 Chr.2: 112233233-112233233 on Build GRCh38
rs538661848 Chr.2: 112233379-112233379 on Build GRCh38
rs764994051 Chr.2: 112233225-112233225 on Build GRCh38
rs200164333 Chr.2: 112233239-112233239 on Build GRCh38
rs759596818 Chr.2: 112233269-112233270 on Build GRCh38
rs143473380 Chr.2: 112233227-112233228 on Build GRCh38
rs769862355 Chr.2: 112233262-112233262 on Build GRCh38
rs748276126 Chr.2: 112233259-112233259 on Build GRCh38
rs1007290780 Chr.2: 112233316-112233316 on Build GRCh38
rs773458669 Chr.2: 112233276-112233276 on Build GRCh38
rs751775476 Chr.2: 112233237-112233237 on Build GRCh38

View Full Product Details