Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113886444-113886717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4B1-AS1
Entrez Gene ID
100287722
Gene Name
AP4B1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.664669
Gene Chromosome Location
Chr.1: 113812612-113901237 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Gene Symbol
BCL2L15
Entrez Gene ID
440603
Gene Name
BCL2 like 15
Gene Aliases
Bfk, C1orf178
UniGene
Hs.123106
Gene Chromosome Location
Chr.1: 113876814-113887547 on Build GRCh38
Transcripts
NR_037864.1 NR_125965.1 NM_001010922.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4533418 c.197G>A p.G66E Chr.1: 113886589-113886589 on Build GRCh38
1332825 c.215C>T p.A72V Chr.1: 113886571-113886571 on Build GRCh38
894086 c.167G>C p.R56P Chr.1: 113886619-113886619 on Build GRCh38
6265465 c.156C>A p.I52I Chr.1: 113886630-113886630 on Build GRCh38
73821 c.193A>C p.N65H Chr.1: 113886593-113886593 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746058780 Chr.1: 113886692-113886692 on Build GRCh38
rs758090026 Chr.1: 113886507-113886507 on Build GRCh38
rs200915499 Chr.1: 113886496-113886496 on Build GRCh38
rs878956070 Chr.1: 113886625-113886625 on Build GRCh38
rs776997453 Chr.1: 113886572-113886572 on Build GRCh38
rs184259183 Chr.1: 113886511-113886511 on Build GRCh38
rs747332311 Chr.1: 113886656-113886656 on Build GRCh38
rs75493886 Chr.1: 113886591-113886591 on Build GRCh38
rs747389971 Chr.1: 113886550-113886550 on Build GRCh38
rs199749160 Chr.1: 113886619-113886619 on Build GRCh38
rs770025111 Chr.1: 113886588-113886588 on Build GRCh38
rs772493129 Chr.1: 113886537-113886537 on Build GRCh38
rs753335741 Chr.1: 113886632-113886632 on Build GRCh38
rs372771274 Chr.1: 113886681-113886681 on Build GRCh38
rs200207977 Chr.1: 113886539-113886539 on Build GRCh38
rs751367925 Chr.1: 113886613-113886613 on Build GRCh38
rs966890474 Chr.1: 113886508-113886508 on Build GRCh38
rs200968967 Chr.1: 113886515-113886515 on Build GRCh38
rs371030564 Chr.1: 113886521-113886521 on Build GRCh38
rs770586700 Chr.1: 113886496-113886496 on Build GRCh38
rs755630175 Chr.1: 113886519-113886519 on Build GRCh38
rs762528661 Chr.1: 113886494-113886494 on Build GRCh38
rs750094333 Chr.1: 113886620-113886620 on Build GRCh38
rs756893176 Chr.1: 113886514-113886514 on Build GRCh38
rs749873143 Chr.1: 113886517-113886517 on Build GRCh38
rs367547795 Chr.1: 113886653-113886653 on Build GRCh38
rs375010375 Chr.1: 113886638-113886638 on Build GRCh38
rs771444622 Chr.1: 113886687-113886687 on Build GRCh38
rs374054881 Chr.1: 113886577-113886577 on Build GRCh38
rs774115399 Chr.1: 113886495-113886496 on Build GRCh38
rs201858571 Chr.1: 113886501-113886501 on Build GRCh38
rs948434236 Chr.1: 113886636-113886636 on Build GRCh38
rs781525849 Chr.1: 113886688-113886688 on Build GRCh38
rs748717712 Chr.1: 113886524-113886524 on Build GRCh38
rs756966006 Chr.1: 113886614-113886614 on Build GRCh38
rs1042722640 Chr.1: 113886663-113886663 on Build GRCh38
rs368105653 Chr.1: 113886615-113886615 on Build GRCh38
rs202118157 Chr.1: 113886628-113886628 on Build GRCh38
rs771272868 Chr.1: 113886557-113886557 on Build GRCh38
rs775474589 Chr.1: 113886590-113886590 on Build GRCh38
rs78559267 Chr.1: 113886626-113886626 on Build GRCh38
rs140487072 Chr.1: 113886512-113886512 on Build GRCh38

View Full Product Details