Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 184245441-184245672 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG3
Entrez Gene ID
10195
Gene Name
ALG3, alpha-1,3- mannosyltransferase
Gene Aliases
CDG1D, CDGS4, CDGS6, D16Ertd36e, NOT56L, Not56, not
UniGene
Hs.478481
Gene Chromosome Location
Chr.3: 184242329-184249525 on Build GRCh38
Transcripts
NM_001006941.2 NM_005787.5 NR_024533.1 NR_024534.1 XM_011512322.1 XM_011512323.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182520 c.403A>G p.T135A Chr.3: 184245509-184245509 on Build GRCh38
6261175 c.201C>A p.T67T Chr.3: 184245567-184245567 on Build GRCh38
7298945 c.313G>A p.V105M Chr.3: 184245599-184245599 on Build GRCh38
4884666 c.205C>T p.R69* Chr.3: 184245563-184245563 on Build GRCh38
6511818 c.221G>T p.R74L Chr.3: 184245547-184245547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs28940588 Chr.3: 184245559-184245559 on Build GRCh38
rs753416504 Chr.3: 184245569-184245569 on Build GRCh38
rs376100127 Chr.3: 184245583-184245583 on Build GRCh38
rs562487441 Chr.3: 184245565-184245565 on Build GRCh38
rs778342644 Chr.3: 184245635-184245635 on Build GRCh38
rs754712887 Chr.3: 184245575-184245576 on Build GRCh38
rs906854585 Chr.3: 184245634-184245634 on Build GRCh38
rs766613289 Chr.3: 184245558-184245558 on Build GRCh38
rs756804378 Chr.3: 184245629-184245629 on Build GRCh38
rs371534425 Chr.3: 184245621-184245621 on Build GRCh38
rs778594200 Chr.3: 184245571-184245571 on Build GRCh38
rs11550775 Chr.3: 184245522-184245522 on Build GRCh38
rs981649329 Chr.3: 184245594-184245594 on Build GRCh38
rs572707309 Chr.3: 184245461-184245461 on Build GRCh38
rs867376445 Chr.3: 184245547-184245547 on Build GRCh38
rs1037322603 Chr.3: 184245468-184245468 on Build GRCh38
rs771885462 Chr.3: 184245576-184245576 on Build GRCh38
rs763104840 Chr.3: 184245548-184245548 on Build GRCh38
rs370434427 Chr.3: 184245562-184245562 on Build GRCh38
rs933979722 Chr.3: 184245537-184245537 on Build GRCh38
rs555083969 Chr.3: 184245513-184245513 on Build GRCh38
rs745555108 Chr.3: 184245574-184245574 on Build GRCh38
rs761211153 Chr.3: 184245611-184245611 on Build GRCh38
rs753238253 Chr.3: 184245563-184245563 on Build GRCh38
rs188264452 Chr.3: 184245497-184245497 on Build GRCh38
rs769858984 Chr.3: 184245543-184245543 on Build GRCh38
rs770957102 Chr.3: 184245587-184245587 on Build GRCh38
rs778518483 Chr.3: 184245487-184245487 on Build GRCh38
rs767923381 Chr.3: 184245600-184245600 on Build GRCh38
rs756927143 Chr.3: 184245570-184245570 on Build GRCh38
rs754433000 Chr.3: 184245626-184245626 on Build GRCh38
rs951800544 Chr.3: 184245553-184245553 on Build GRCh38
rs746880853 Chr.3: 184245577-184245577 on Build GRCh38
rs753012485 Chr.3: 184245602-184245602 on Build GRCh38
rs760134525 Chr.3: 184245516-184245516 on Build GRCh38
rs140738312 Chr.3: 184245536-184245536 on Build GRCh38
rs770718666 Chr.3: 184245480-184245480 on Build GRCh38
rs759737007 Chr.3: 184245595-184245595 on Build GRCh38
rs377372523 Chr.3: 184245545-184245545 on Build GRCh38
rs773227426 Chr.3: 184245580-184245580 on Build GRCh38
rs748878963 Chr.3: 184245467-184245467 on Build GRCh38
rs745702853 Chr.3: 184245503-184245503 on Build GRCh38
rs991645427 Chr.3: 184245627-184245627 on Build GRCh38
rs2233463 Chr.3: 184245593-184245593 on Build GRCh38

View Full Product Details