Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 105552573-105552820 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF38
Entrez Gene ID
54848
Gene Name
Rho guanine nucleotide exchange factor 38
Gene Aliases
-
UniGene
Hs.585224
Gene Chromosome Location
Chr.4: 105552603-105682539 on Build GRCh38
Transcripts
NM_001242729.1 NM_017700.1 XM_011532051.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1594200 c.13G>T p.E5* Chr.4: 105552778-105552778 on Build GRCh38
6604312 c.39C>T p.V13V Chr.4: 105552804-105552804 on Build GRCh38
1049776 c.13G>T p.E5* Chr.4: 105552778-105552778 on Build GRCh38
1642376 c.32A>C p.N11T Chr.4: 105552797-105552797 on Build GRCh38
6604311 c.39C>T p.V13V Chr.4: 105552804-105552804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112195119 Chr.4: 105552642-105552642 on Build GRCh38
rs757174787 Chr.4: 105552765-105552765 on Build GRCh38
rs572294831 Chr.4: 105552679-105552679 on Build GRCh38
rs755925399 Chr.4: 105552718-105552718 on Build GRCh38
rs1002942686 Chr.4: 105552606-105552606 on Build GRCh38
rs779085578 Chr.4: 105552778-105552778 on Build GRCh38
rs965565840 Chr.4: 105552780-105552780 on Build GRCh38
rs752417042 Chr.4: 105552715-105552715 on Build GRCh38
rs749184437 Chr.4: 105552763-105552763 on Build GRCh38
rs745919785 Chr.4: 105552786-105552786 on Build GRCh38
rs758224060 Chr.4: 105552612-105552612 on Build GRCh38
rs559868186 Chr.4: 105552637-105552637 on Build GRCh38
rs75086757 Chr.4: 105552705-105552705 on Build GRCh38
rs754455990 Chr.4: 105552714-105552714 on Build GRCh38
rs897141518 Chr.4: 105552650-105552650 on Build GRCh38
rs1019776161 Chr.4: 105552730-105552730 on Build GRCh38
rs1027618260 Chr.4: 105552699-105552699 on Build GRCh38
rs562263895 Chr.4: 105552767-105552767 on Build GRCh38
rs772317417 Chr.4: 105552791-105552791 on Build GRCh38
rs777443919 Chr.4: 105552720-105552720 on Build GRCh38
rs1034056168 Chr.4: 105552627-105552627 on Build GRCh38

View Full Product Details