Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112163379-112163606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD4
Entrez Gene ID
283450
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Gene Aliases
C12orf51, POTAGE
UniGene
Hs.530943
Gene Chromosome Location
Chr.12: 112160188-112382092 on Build GRCh38
Transcripts
NM_001109662.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3954189 c.11606C>T p.P3869L Chr.12: 112163567-112163567 on Build GRCh38
4448753 c.11631+2T>C p.? Chr.12: 112163540-112163540 on Build GRCh38
4652137 c.11590C>T p.R3864C Chr.12: 112163583-112163583 on Build GRCh38
7228027 c.11606C>G p.P3869R Chr.12: 112163567-112163567 on Build GRCh38
5583318 c.11619C>T p.L3873L Chr.12: 112163554-112163554 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs2272308 Chr.12: 112163446-112163446 on Build GRCh38
rs114947184 Chr.12: 112163437-112163437 on Build GRCh38
rs566632838 Chr.12: 112163455-112163455 on Build GRCh38
rs939560243 Chr.12: 112163456-112163456 on Build GRCh38
rs534136597 Chr.12: 112163512-112163512 on Build GRCh38
rs1002601412 Chr.12: 112163522-112163522 on Build GRCh38
rs1048043143 Chr.12: 112163500-112163500 on Build GRCh38
rs988684909 Chr.12: 112163564-112163564 on Build GRCh38
rs35084322 Chr.12: 112163530-112163531 on Build GRCh38
rs1022679472 Chr.12: 112163516-112163516 on Build GRCh38
rs929533406 Chr.12: 112163494-112163494 on Build GRCh38
rs1035473318 Chr.12: 112163548-112163548 on Build GRCh38
rs775441229 Chr.12: 112163588-112163588 on Build GRCh38
rs1040585541 Chr.12: 112163487-112163487 on Build GRCh38
rs887449700 Chr.12: 112163505-112163505 on Build GRCh38
rs775792386 Chr.12: 112163515-112163515 on Build GRCh38
rs573784336 Chr.12: 112163575-112163575 on Build GRCh38
rs77746486 Chr.12: 112163563-112163563 on Build GRCh38
rs375938583 Chr.12: 112163551-112163551 on Build GRCh38
rs902038325 Chr.12: 112163493-112163493 on Build GRCh38
rs771253864 Chr.12: 112163517-112163517 on Build GRCh38

View Full Product Details