Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 29329476-29329701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BACH1
Entrez Gene ID
571
Gene Name
BTB domain and CNC homolog 1
Gene Aliases
BACH-1, BTBD24
UniGene
Hs.154276
Gene Chromosome Location
Chr.21: 29298795-29361896 on Build GRCh38
Transcripts
NM_001186.3 NM_206866.2 NR_027655.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3771127 c.1700A>G p.N567S Chr.21: 29329617-29329617 on Build GRCh38
324792 c.1624C>T p.Q542* Chr.21: 29329541-29329541 on Build GRCh38
1184357 c.1586A>G p.N529S Chr.21: 29329503-29329503 on Build GRCh38
419276 c.1624C>G p.Q542E Chr.21: 29329541-29329541 on Build GRCh38
2841860 c.1672A>G p.I558V Chr.21: 29329589-29329589 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749530429 Chr.21: 29329599-29329599 on Build GRCh38
rs774134757 Chr.21: 29329611-29329611 on Build GRCh38
rs752226287 Chr.21: 29329511-29329511 on Build GRCh38
rs761591200 Chr.21: 29329661-29329661 on Build GRCh38
rs751969325 Chr.21: 29329672-29329672 on Build GRCh38
rs950915119 Chr.21: 29329609-29329609 on Build GRCh38
rs925316451 Chr.21: 29329607-29329607 on Build GRCh38
rs776374291 Chr.21: 29329640-29329640 on Build GRCh38
rs770670228 Chr.21: 29329602-29329602 on Build GRCh38
rs760229936 Chr.21: 29329528-29329528 on Build GRCh38
rs532626668 Chr.21: 29329563-29329563 on Build GRCh38
rs758884168 Chr.21: 29329543-29329543 on Build GRCh38
rs766536082 Chr.21: 29329602-29329604 on Build GRCh38
rs776875376 Chr.21: 29329641-29329642 on Build GRCh38
rs745733026 Chr.21: 29329620-29329620 on Build GRCh38
rs750785692 Chr.21: 29329530-29329530 on Build GRCh38
rs772104130 Chr.21: 29329627-29329627 on Build GRCh38
rs760272033 Chr.21: 29329634-29329634 on Build GRCh38
rs766868523 Chr.21: 29329515-29329515 on Build GRCh38
rs752073995 Chr.21: 29329547-29329547 on Build GRCh38
rs766873491 Chr.21: 29329545-29329545 on Build GRCh38
rs775077415 Chr.21: 29329524-29329524 on Build GRCh38
rs763873552 Chr.21: 29329529-29329529 on Build GRCh38
rs140484641 Chr.21: 29329635-29329635 on Build GRCh38
rs935323231 Chr.21: 29329628-29329628 on Build GRCh38
rs755516583 Chr.21: 29329558-29329558 on Build GRCh38
rs375476274 Chr.21: 29329560-29329560 on Build GRCh38
rs766784816 Chr.21: 29329668-29329668 on Build GRCh38
rs774857095 Chr.21: 29329629-29329629 on Build GRCh38
rs1044331216 Chr.21: 29329512-29329512 on Build GRCh38
rs778014862 Chr.21: 29329568-29329568 on Build GRCh38
rs773460146 Chr.21: 29329612-29329613 on Build GRCh38
rs755430603 Chr.21: 29329674-29329674 on Build GRCh38

View Full Product Details