Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 33867656-33867901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AVEN
Entrez Gene ID
57099
Gene Name
apoptosis and caspase activation inhibitor
Gene Aliases
PDCD12
UniGene
Hs.555966
Gene Chromosome Location
Chr.15: 33858602-34074877 on Build GRCh38
Transcripts
NM_020371.2 XM_011521818.2 XM_011521819.2 XM_011521820.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5906408 c.719C>T p.P240L Chr.15: 33867749-33867749 on Build GRCh38
5806914 c.706G>T p.G236* Chr.15: 33867762-33867762 on Build GRCh38
3744557 c.744T>C p.A248A Chr.15: 33867724-33867724 on Build GRCh38
4505578 c.696C>T p.P232P Chr.15: 33867772-33867772 on Build GRCh38
6521962 c.707G>T p.G236V Chr.15: 33867761-33867761 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370073555 Chr.15: 33867704-33867704 on Build GRCh38
rs61729120 Chr.15: 33867768-33867768 on Build GRCh38
rs368260695 Chr.15: 33867821-33867821 on Build GRCh38
rs747539138 Chr.15: 33867812-33867812 on Build GRCh38
rs772687615 Chr.15: 33867713-33867713 on Build GRCh38
rs752587332 Chr.15: 33867778-33867778 on Build GRCh38
rs751557522 Chr.15: 33867682-33867682 on Build GRCh38
rs148454789 Chr.15: 33867851-33867851 on Build GRCh38
rs932439173 Chr.15: 33867827-33867827 on Build GRCh38
rs369651698 Chr.15: 33867795-33867795 on Build GRCh38
rs148136950 Chr.15: 33867693-33867693 on Build GRCh38
rs370527932 Chr.15: 33867715-33867715 on Build GRCh38
rs746419966 Chr.15: 33867710-33867710 on Build GRCh38
rs766230719 Chr.15: 33867733-33867733 on Build GRCh38
rs776096451 Chr.15: 33867714-33867714 on Build GRCh38
rs748202987 Chr.15: 33867691-33867691 on Build GRCh38
rs757494978 Chr.15: 33867751-33867751 on Build GRCh38
rs770388146 Chr.15: 33867833-33867833 on Build GRCh38
rs916945049 Chr.15: 33867772-33867772 on Build GRCh38
rs751932445 Chr.15: 33867870-33867870 on Build GRCh38
rs774405935 Chr.15: 33867715-33867721 on Build GRCh38
rs746260258 Chr.15: 33867753-33867753 on Build GRCh38
rs2241647 Chr.15: 33867785-33867785 on Build GRCh38
rs1029464028 Chr.15: 33867711-33867711 on Build GRCh38
rs962636297 Chr.15: 33867750-33867751 on Build GRCh38
rs759357755 Chr.15: 33867775-33867775 on Build GRCh38
rs564664081 Chr.15: 33867683-33867683 on Build GRCh38
rs149958275 Chr.15: 33867786-33867786 on Build GRCh38
rs149320627 Chr.15: 33867794-33867794 on Build GRCh38
rs376955952 Chr.15: 33867699-33867699 on Build GRCh38
rs532118544 Chr.15: 33867745-33867745 on Build GRCh38
rs777847400 Chr.15: 33867748-33867748 on Build GRCh38
rs759127003 Chr.15: 33867842-33867842 on Build GRCh38
rs775349309 Chr.15: 33867846-33867846 on Build GRCh38
rs777887424 Chr.15: 33867695-33867695 on Build GRCh38
rs769246123 Chr.15: 33867762-33867762 on Build GRCh38
rs374878529 Chr.15: 33867776-33867776 on Build GRCh38
rs144112753 Chr.15: 33867869-33867869 on Build GRCh38
rs749456950 Chr.15: 33867696-33867696 on Build GRCh38
rs137887529 Chr.15: 33867761-33867761 on Build GRCh38
rs1049153621 Chr.15: 33867790-33867790 on Build GRCh38
rs757238065 Chr.15: 33867787-33867787 on Build GRCh38
rs762656596 Chr.15: 33867731-33867731 on Build GRCh38
rs750609985 Chr.15: 33867792-33867792 on Build GRCh38
rs752367062 Chr.15: 33867766-33867769 on Build GRCh38
rs765243912 Chr.15: 33867705-33867705 on Build GRCh38
rs61729119 Chr.15: 33867740-33867740 on Build GRCh38
rs760645906 Chr.15: 33867779-33867779 on Build GRCh38
rs765353935 Chr.15: 33867867-33867867 on Build GRCh38
rs781288336 Chr.15: 33867689-33867689 on Build GRCh38
rs1009275017 Chr.15: 33867729-33867729 on Build GRCh38
rs771908835 Chr.15: 33867845-33867845 on Build GRCh38
rs759306134 Chr.15: 33867751-33867751 on Build GRCh38
rs760398550 Chr.15: 33867771-33867772 on Build GRCh38
rs779312581 Chr.15: 33867791-33867791 on Build GRCh38
rs201675744 Chr.15: 33867805-33867805 on Build GRCh38
rs767338315 Chr.15: 33867757-33867758 on Build GRCh38
rs777202554 Chr.15: 33867725-33867725 on Build GRCh38
rs751322513 Chr.15: 33867735-33867735 on Build GRCh38
rs763697165 Chr.15: 33867790-33867791 on Build GRCh38
rs372220481 Chr.15: 33867822-33867822 on Build GRCh38
rs749529048 Chr.15: 33867749-33867749 on Build GRCh38
rs772754452 Chr.15: 33867726-33867726 on Build GRCh38
rs779472746 Chr.15: 33867752-33867752 on Build GRCh38
rs141686976 Chr.15: 33867721-33867721 on Build GRCh38
rs772530130 Chr.15: 33867756-33867756 on Build GRCh38
rs116106056 Chr.15: 33867830-33867830 on Build GRCh38
rs773943038 Chr.15: 33867773-33867773 on Build GRCh38
rs552179779 Chr.15: 33867862-33867862 on Build GRCh38

View Full Product Details