Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 69084955-69085186 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL6IP5
Entrez Gene ID
10550
Gene Name
ADP ribosylation factor like GTPase 6 interacting protein 5
Gene Aliases
DERP11, GTRAP3-18, HSPC127, JWA, PRAF3, Yip6b, addicsin, hp22, jmx
UniGene
Hs.518060
Gene Chromosome Location
Chr.3: 69084939-69106088 on Build GRCh38
Transcripts
NM_006407.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4845808 c.15C>G p.I5M Chr.3: 69085062-69085062 on Build GRCh38
7200614 c.41T>G p.F14C Chr.3: 69085088-69085088 on Build GRCh38
7094897 c.42C>A p.F14L Chr.3: 69085089-69085089 on Build GRCh38
4158309 c.22C>T p.L8F Chr.3: 69085069-69085069 on Build GRCh38
1753377 c.4G>A p.D2N Chr.3: 69085051-69085051 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs528001807 Chr.3: 69085075-69085075 on Build GRCh38
rs899160016 Chr.3: 69085118-69085118 on Build GRCh38
rs767557247 Chr.3: 69084998-69084999 on Build GRCh38
rs751081080 Chr.3: 69085008-69085008 on Build GRCh38
rs772673187 Chr.3: 69085102-69085102 on Build GRCh38
rs774102206 Chr.3: 69085153-69085153 on Build GRCh38
rs111700508 Chr.3: 69085033-69085033 on Build GRCh38
rs149212853 Chr.3: 69085007-69085007 on Build GRCh38
rs751545675 Chr.3: 69085151-69085151 on Build GRCh38
rs11542220 Chr.3: 69085137-69085137 on Build GRCh38
rs762750764 Chr.3: 69085048-69085048 on Build GRCh38
rs201200564 Chr.3: 69085114-69085114 on Build GRCh38
rs888595897 Chr.3: 69084995-69084996 on Build GRCh38
rs767055034 Chr.3: 69085053-69085053 on Build GRCh38
rs947976176 Chr.3: 69084980-69084980 on Build GRCh38
rs779700699 Chr.3: 69085076-69085076 on Build GRCh38
rs779589390 Chr.3: 69085094-69085094 on Build GRCh38
rs202151518 Chr.3: 69085030-69085030 on Build GRCh38
rs780762487 Chr.3: 69084996-69084998 on Build GRCh38
rs758331284 Chr.3: 69085001-69085001 on Build GRCh38
rs747095530 Chr.3: 69085111-69085111 on Build GRCh38
rs372636665 Chr.3: 69085093-69085093 on Build GRCh38
rs906410779 Chr.3: 69084991-69084991 on Build GRCh38
rs768390281 Chr.3: 69085139-69085139 on Build GRCh38
rs1016275373 Chr.3: 69085072-69085072 on Build GRCh38
rs989176555 Chr.3: 69084975-69084975 on Build GRCh38
rs569349303 Chr.3: 69084979-69084979 on Build GRCh38
rs557684428 Chr.3: 69085082-69085082 on Build GRCh38
rs748273789 Chr.3: 69085035-69085035 on Build GRCh38
rs774935049 Chr.3: 69085034-69085034 on Build GRCh38
rs534878576 Chr.3: 69085067-69085067 on Build GRCh38
rs764022062 Chr.3: 69085049-69085049 on Build GRCh38
rs745570847 Chr.3: 69085026-69085026 on Build GRCh38
rs780853153 Chr.3: 69085025-69085025 on Build GRCh38
rs749621819 Chr.3: 69085077-69085077 on Build GRCh38
rs775204291 Chr.3: 69085044-69085044 on Build GRCh38
rs755761324 Chr.3: 69085027-69085027 on Build GRCh38
rs1044958983 Chr.3: 69084986-69084986 on Build GRCh38
rs756689357 Chr.3: 69085010-69085010 on Build GRCh38
rs749945961 Chr.3: 69085058-69085058 on Build GRCh38
rs776844556 Chr.3: 69085112-69085112 on Build GRCh38
rs903202730 Chr.3: 69085107-69085107 on Build GRCh38
rs368886970 Chr.3: 69085036-69085036 on Build GRCh38
rs766616578 Chr.3: 69085037-69085037 on Build GRCh38
rs376490250 Chr.3: 69085104-69085104 on Build GRCh38

View Full Product Details