Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 31983126-31983385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1551
Entrez Gene ID
55196
Gene Name
KIAA1551
Gene Aliases
C12orf35, UTA2-1
UniGene
Hs.445129
Gene Chromosome Location
Chr.12: 31958800-31993107 on Build GRCh38
Transcripts
NM_018169.3 XM_005253405.2 XM_006719099.2 XM_011520722.1 XM_017019550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1163001 c.2286A>G p.V762V Chr.12: 31983241-31983241 on Build GRCh38
5751005 c.2294C>A p.P765Q Chr.12: 31983249-31983249 on Build GRCh38
6349968 c.2333G>A p.G778E Chr.12: 31983288-31983288 on Build GRCh38
938777 c.2223A>G p.V741V Chr.12: 31983178-31983178 on Build GRCh38
6585690 c.2246G>A p.G749D Chr.12: 31983201-31983201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773331894 Chr.12: 31983302-31983302 on Build GRCh38
rs139342835 Chr.12: 31983288-31983288 on Build GRCh38
rs768682003 Chr.12: 31983249-31983249 on Build GRCh38
rs917583661 Chr.12: 31983212-31983212 on Build GRCh38
rs148931564 Chr.12: 31983269-31983269 on Build GRCh38
rs1004372647 Chr.12: 31983198-31983198 on Build GRCh38
rs529070440 Chr.12: 31983290-31983290 on Build GRCh38
rs769274490 Chr.12: 31983163-31983163 on Build GRCh38
rs772537490 Chr.12: 31983300-31983300 on Build GRCh38
rs774658680 Chr.12: 31983165-31983165 on Build GRCh38
rs1019655398 Chr.12: 31983210-31983210 on Build GRCh38
rs558126712 Chr.12: 31983364-31983364 on Build GRCh38
rs780513294 Chr.12: 31983160-31983160 on Build GRCh38
rs763753313 Chr.12: 31983326-31983326 on Build GRCh38
rs751491697 Chr.12: 31983216-31983216 on Build GRCh38
rs965395459 Chr.12: 31983268-31983268 on Build GRCh38
rs755885585 Chr.12: 31983251-31983251 on Build GRCh38
rs143720733 Chr.12: 31983208-31983208 on Build GRCh38
rs145743642 Chr.12: 31983254-31983254 on Build GRCh38
rs771188313 Chr.12: 31983321-31983321 on Build GRCh38
rs749606710 Chr.12: 31983161-31983161 on Build GRCh38
rs767171915 Chr.12: 31983241-31983241 on Build GRCh38
rs201519644 Chr.12: 31983298-31983298 on Build GRCh38
rs761887952 Chr.12: 31983352-31983352 on Build GRCh38
rs534318377 Chr.12: 31983250-31983250 on Build GRCh38
rs774252528 Chr.12: 31983347-31983347 on Build GRCh38
rs908986439 Chr.12: 31983267-31983267 on Build GRCh38
rs760593097 Chr.12: 31983199-31983199 on Build GRCh38
rs369502968 Chr.12: 31983190-31983190 on Build GRCh38
rs533047029 Chr.12: 31983166-31983166 on Build GRCh38
rs887270296 Chr.12: 31983197-31983197 on Build GRCh38
rs756855377 Chr.12: 31983232-31983232 on Build GRCh38
rs767546025 Chr.12: 31983353-31983353 on Build GRCh38
rs779571886 Chr.12: 31983265-31983265 on Build GRCh38
rs546535389 Chr.12: 31983186-31983186 on Build GRCh38
rs773454747 Chr.12: 31983172-31983172 on Build GRCh38
rs763354998 Chr.12: 31983197-31983198 on Build GRCh38
rs750023477 Chr.12: 31983356-31983356 on Build GRCh38
rs1045124921 Chr.12: 31983257-31983257 on Build GRCh38
rs764106305 Chr.12: 31983213-31983213 on Build GRCh38
rs537814008 Chr.12: 31983325-31983325 on Build GRCh38
rs143630974 Chr.12: 31983317-31983317 on Build GRCh38
rs772091138 Chr.12: 31983171-31983171 on Build GRCh38

View Full Product Details