Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94639168-94639397 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBP1
Entrez Gene ID
2203
Gene Name
fructose-bisphosphatase 1
Gene Aliases
FBP
UniGene
Hs.494496
Gene Chromosome Location
Chr.9: 94603133-94640257 on Build GRCh38
Transcripts
NM_000507.3 NM_001127628.1 XM_006717005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3413855 c.45C>G p.T15T Chr.9: 94639266-94639266 on Build GRCh38
7287257 c.4G>A p.A2T Chr.9: 94639307-94639307 on Build GRCh38
5504988 c.51C>T p.F17F Chr.9: 94639260-94639260 on Build GRCh38
1463775 c.88G>A p.E30K Chr.9: 94639223-94639223 on Build GRCh38
4569766 c.19T>C p.F7L Chr.9: 94639292-94639292 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758509921 Chr.9: 94639329-94639355 on Build GRCh38
rs773395852 Chr.9: 94639293-94639293 on Build GRCh38
rs767460321 Chr.9: 94639230-94639230 on Build GRCh38
rs143832764 Chr.9: 94639209-94639209 on Build GRCh38
rs769000568 Chr.9: 94639272-94639272 on Build GRCh38
rs542675680 Chr.9: 94639332-94639332 on Build GRCh38
rs121918190 Chr.9: 94639223-94639223 on Build GRCh38
rs945416489 Chr.9: 94639374-94639374 on Build GRCh38
rs200720843 Chr.9: 94639264-94639264 on Build GRCh38
rs958446114 Chr.9: 94639337-94639337 on Build GRCh38
rs774456590 Chr.9: 94639227-94639227 on Build GRCh38
rs759615686 Chr.9: 94639297-94639297 on Build GRCh38
rs867795895 Chr.9: 94639232-94639232 on Build GRCh38
rs997609658 Chr.9: 94639298-94639298 on Build GRCh38
rs957149631 Chr.9: 94639375-94639375 on Build GRCh38
rs796383791 Chr.9: 94639263-94639263 on Build GRCh38
rs747358501 Chr.9: 94639271-94639271 on Build GRCh38
rs776753141 Chr.9: 94639278-94639278 on Build GRCh38
rs746418821 Chr.9: 94639261-94639261 on Build GRCh38
rs748356162 Chr.9: 94639284-94639284 on Build GRCh38
rs759527980 Chr.9: 94639229-94639229 on Build GRCh38
rs796214094 Chr.9: 94639259-94639260 on Build GRCh38
rs578183426 Chr.9: 94639248-94639248 on Build GRCh38
rs898005350 Chr.9: 94639275-94639275 on Build GRCh38
rs1057515691 Chr.9: 94639369-94639369 on Build GRCh38
rs752128198 Chr.9: 94639349-94639350 on Build GRCh38
rs747477454 Chr.9: 94639333-94639333 on Build GRCh38
rs749394622 Chr.9: 94639361-94639361 on Build GRCh38
rs756749421 Chr.9: 94639254-94639254 on Build GRCh38
rs770051809 Chr.9: 94639358-94639358 on Build GRCh38
rs753462820 Chr.9: 94639251-94639251 on Build GRCh38
rs752536186 Chr.9: 94639239-94639239 on Build GRCh38
rs775512212 Chr.9: 94639306-94639306 on Build GRCh38
rs760389518 Chr.9: 94639318-94639318 on Build GRCh38
rs1057515690 Chr.9: 94639364-94639364 on Build GRCh38
rs778057537 Chr.9: 94639359-94639359 on Build GRCh38
rs767550290 Chr.9: 94639300-94639300 on Build GRCh38
rs369411593 Chr.9: 94639319-94639319 on Build GRCh38
rs572096460 Chr.9: 94639331-94639331 on Build GRCh38
rs764449536 Chr.9: 94639346-94639347 on Build GRCh38
rs913996916 Chr.9: 94639369-94639369 on Build GRCh38
rs1017934707 Chr.9: 94639342-94639342 on Build GRCh38
rs748363187 Chr.9: 94639355-94639355 on Build GRCh38
rs769817624 Chr.9: 94639289-94639289 on Build GRCh38
rs781602148 Chr.9: 94639351-94639351 on Build GRCh38
rs545695727 Chr.9: 94639260-94639260 on Build GRCh38
rs749926444 Chr.9: 94639329-94639329 on Build GRCh38
rs765322852 Chr.9: 94639215-94639215 on Build GRCh38
rs556756210 Chr.9: 94639245-94639245 on Build GRCh38
rs1006994685 Chr.9: 94639328-94639328 on Build GRCh38
rs755324351 Chr.9: 94639348-94639348 on Build GRCh38
rs199970657 Chr.9: 94639320-94639320 on Build GRCh38

View Full Product Details