Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120404772-120405045 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL1
Entrez Gene ID
11167
Gene Name
follistatin like 1
Gene Aliases
FRP, FSL1, MIR198, OCC-1, OCC1, tsc36
UniGene
Hs.269512
Gene Chromosome Location
Chr.3: 120394214-120451071 on Build GRCh38
Transcripts
NM_007085.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1036856 c.578T>G p.L193R Chr.3: 120404856-120404856 on Build GRCh38
3585826 c.537C>T p.I179I Chr.3: 120404897-120404897 on Build GRCh38
5974495 c.581+1G>A p.? Chr.3: 120404852-120404852 on Build GRCh38
1036858 c.501A>G p.E167E Chr.3: 120404933-120404933 on Build GRCh38
1036857 c.549G>T p.T183T Chr.3: 120404885-120404885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150197564 Chr.3: 120404950-120404950 on Build GRCh38
rs759840922 Chr.3: 120404983-120404983 on Build GRCh38
rs138829728 Chr.3: 120404885-120404885 on Build GRCh38
rs779591019 Chr.3: 120404952-120404952 on Build GRCh38
rs757655104 Chr.3: 120404935-120404935 on Build GRCh38
rs1028889387 Chr.3: 120404896-120404896 on Build GRCh38
rs769256525 Chr.3: 120404827-120404827 on Build GRCh38
rs571541119 Chr.3: 120404998-120404998 on Build GRCh38
rs776677434 Chr.3: 120404980-120404980 on Build GRCh38
rs770558639 Chr.3: 120404817-120404817 on Build GRCh38
rs765384446 Chr.3: 120404987-120404987 on Build GRCh38
rs751806112 Chr.3: 120404933-120404933 on Build GRCh38
rs555454416 Chr.3: 120405009-120405009 on Build GRCh38
rs374027382 Chr.3: 120404850-120404850 on Build GRCh38
rs749212088 Chr.3: 120404958-120404958 on Build GRCh38
rs148160939 Chr.3: 120404867-120404867 on Build GRCh38
rs557810477 Chr.3: 120404994-120404994 on Build GRCh38
rs745312872 Chr.3: 120404825-120404825 on Build GRCh38
rs757526039 Chr.3: 120405000-120405000 on Build GRCh38
rs752881149 Chr.3: 120404993-120404993 on Build GRCh38
rs1000403177 Chr.3: 120404797-120404797 on Build GRCh38
rs746649202 Chr.3: 120404804-120404804 on Build GRCh38
rs775367384 Chr.3: 120404840-120404840 on Build GRCh38
rs745569949 Chr.3: 120404949-120404949 on Build GRCh38
rs183355920 Chr.3: 120404892-120404892 on Build GRCh38
rs371335574 Chr.3: 120404895-120404895 on Build GRCh38
rs780659891 Chr.3: 120404936-120404936 on Build GRCh38
rs776354559 Chr.3: 120404822-120404822 on Build GRCh38
rs751235378 Chr.3: 120404862-120404864 on Build GRCh38
rs140954100 Chr.3: 120404910-120404910 on Build GRCh38
rs774187993 Chr.3: 120404862-120404862 on Build GRCh38
rs764638612 Chr.3: 120404997-120404997 on Build GRCh38
rs374264814 Chr.3: 120404930-120404930 on Build GRCh38
rs759485845 Chr.3: 120404876-120404876 on Build GRCh38
rs781653528 Chr.3: 120405003-120405003 on Build GRCh38
rs772823456 Chr.3: 120404841-120404841 on Build GRCh38
rs768644365 Chr.3: 120404959-120404959 on Build GRCh38
rs771918360 Chr.3: 120404977-120404977 on Build GRCh38
rs774273723 Chr.3: 120404962-120404962 on Build GRCh38
rs768419039 Chr.3: 120404861-120404861 on Build GRCh38
rs191192598 Chr.3: 120404875-120404875 on Build GRCh38
rs201032578 Chr.3: 120404882-120404882 on Build GRCh38
rs761817450 Chr.3: 120405022-120405032 on Build GRCh38
rs1021955074 Chr.3: 120404886-120404886 on Build GRCh38
rs753231589 Chr.3: 120405005-120405005 on Build GRCh38
rs199863523 Chr.3: 120404971-120404971 on Build GRCh38
rs761373864 Chr.3: 120404793-120404793 on Build GRCh38

View Full Product Details