Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26182196-26182461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNKSR1
Entrez Gene ID
10256
Gene Name
connector enhancer of kinase suppressor of Ras 1
Gene Aliases
CNK, CNK1
UniGene
Hs.16232
Gene Chromosome Location
Chr.1: 26177490-26189884 on Build GRCh38
Transcripts
NM_001297647.1 NM_001297648.1 NM_006314.2 XM_011540484.2 XM_011540485.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3487445 c.502G>A p.G168S Chr.1: 26182385-26182385 on Build GRCh38
6028851 c.519+7C>T p.? Chr.1: 26182409-26182409 on Build GRCh38
4673100 c.488C>T p.A163V Chr.1: 26182371-26182371 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs545535415 Chr.1: 26182275-26182275 on Build GRCh38
rs1004056326 Chr.1: 26182292-26182292 on Build GRCh38
rs776223709 Chr.1: 26182435-26182435 on Build GRCh38
rs919283482 Chr.1: 26182287-26182287 on Build GRCh38
rs760115070 Chr.1: 26182403-26182404 on Build GRCh38
rs749278209 Chr.1: 26182319-26182319 on Build GRCh38
rs191664069 Chr.1: 26182311-26182311 on Build GRCh38
rs751828242 Chr.1: 26182403-26182403 on Build GRCh38
rs1038644416 Chr.1: 26182299-26182299 on Build GRCh38
rs376106983 Chr.1: 26182440-26182440 on Build GRCh38
rs779058664 Chr.1: 26182437-26182437 on Build GRCh38
rs143057209 Chr.1: 26182402-26182402 on Build GRCh38
rs757298491 Chr.1: 26182323-26182323 on Build GRCh38
rs1007861774 Chr.1: 26182336-26182336 on Build GRCh38
rs778969076 Chr.1: 26182331-26182331 on Build GRCh38
rs769100324 Chr.1: 26182366-26182366 on Build GRCh38
rs371649956 Chr.1: 26182293-26182293 on Build GRCh38
rs148174101 Chr.1: 26182371-26182371 on Build GRCh38
rs771700066 Chr.1: 26182333-26182333 on Build GRCh38
rs188766116 Chr.1: 26182276-26182276 on Build GRCh38
rs755115754 Chr.1: 26182415-26182415 on Build GRCh38
rs575660445 Chr.1: 26182283-26182283 on Build GRCh38
rs369629366 Chr.1: 26182315-26182315 on Build GRCh38
rs147255718 Chr.1: 26182381-26182381 on Build GRCh38
rs746597281 Chr.1: 26182364-26182364 on Build GRCh38
rs373210180 Chr.1: 26182430-26182430 on Build GRCh38
rs981686371 Chr.1: 26182249-26182250 on Build GRCh38
rs745425891 Chr.1: 26182332-26182332 on Build GRCh38
rs558089457 Chr.1: 26182222-26182222 on Build GRCh38
rs902086036 Chr.1: 26182374-26182374 on Build GRCh38
rs576834014 Chr.1: 26182244-26182244 on Build GRCh38
rs768194590 Chr.1: 26182418-26182418 on Build GRCh38
rs745729758 Chr.1: 26182438-26182438 on Build GRCh38
rs376576801 Chr.1: 26182313-26182313 on Build GRCh38
rs767746729 Chr.1: 26182418-26182418 on Build GRCh38
rs774898200 Chr.1: 26182327-26182327 on Build GRCh38
rs943510112 Chr.1: 26182350-26182350 on Build GRCh38
rs1027461399 Chr.1: 26182432-26182459 on Build GRCh38
rs763026660 Chr.1: 26182399-26182399 on Build GRCh38
rs765794112 Chr.1: 26182377-26182377 on Build GRCh38
rs775208134 Chr.1: 26182334-26182334 on Build GRCh38
rs373991299 Chr.1: 26182373-26182373 on Build GRCh38
rs779395714 Chr.1: 26182441-26182441 on Build GRCh38
rs757102140 Chr.1: 26182432-26182432 on Build GRCh38
rs950025357 Chr.1: 26182218-26182218 on Build GRCh38

View Full Product Details