Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 35471959-35472232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP250
Entrez Gene ID
11190
Gene Name
centrosomal protein 250
Gene Aliases
C-NAP1, CEP2, CNAP1
UniGene
Hs.443976
Gene Chromosome Location
Chr.20: 35455138-35517531 on Build GRCh38
Transcripts
NM_001318219.1 NM_007186.5 XM_005260262.4 XM_005260263.4 XM_005260264.4 XM_006723690.3 XM_006723691.1 XM_006723692.3 XM_006723693.3 XM_006723694.3 XM_011528517.2 XM_011528518.2 XM_011528519.2 XM_017027617.1 XM_017027618.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6159878 c.951G>C p.E317D Chr.20: 35472052-35472052 on Build GRCh38
3972531 c.987T>C p.S329S Chr.20: 35472088-35472088 on Build GRCh38
459768 c.1035C>G p.I345M Chr.20: 35472136-35472136 on Build GRCh38
4097907 c.1001C>T p.A334V Chr.20: 35472102-35472102 on Build GRCh38
6050608 c.955A>G p.N319D Chr.20: 35472056-35472056 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768722631 Chr.20: 35472044-35472044 on Build GRCh38
rs749323931 Chr.20: 35472019-35472019 on Build GRCh38
rs376261626 Chr.20: 35472170-35472170 on Build GRCh38
rs553115275 Chr.20: 35472193-35472193 on Build GRCh38
rs369255096 Chr.20: 35472110-35472110 on Build GRCh38
rs755080555 Chr.20: 35472182-35472182 on Build GRCh38
rs759362859 Chr.20: 35472158-35472158 on Build GRCh38
rs778930748 Chr.20: 35472183-35472183 on Build GRCh38
rs376259700 Chr.20: 35472106-35472106 on Build GRCh38
rs369235134 Chr.20: 35472103-35472103 on Build GRCh38
rs889288195 Chr.20: 35472177-35472177 on Build GRCh38
rs534828286 Chr.20: 35472169-35472169 on Build GRCh38
rs150639987 Chr.20: 35472083-35472083 on Build GRCh38
rs767701253 Chr.20: 35472031-35472032 on Build GRCh38
rs768045079 Chr.20: 35472167-35472167 on Build GRCh38
rs757809499 Chr.20: 35472166-35472166 on Build GRCh38
rs774294173 Chr.20: 35472062-35472062 on Build GRCh38
rs111334264 Chr.20: 35472051-35472051 on Build GRCh38
rs773626726 Chr.20: 35472107-35472109 on Build GRCh38
rs758986719 Chr.20: 35472128-35472128 on Build GRCh38
rs752145543 Chr.20: 35472161-35472161 on Build GRCh38
rs749477430 Chr.20: 35472180-35472180 on Build GRCh38
rs116166982 Chr.20: 35472102-35472102 on Build GRCh38
rs780152139 Chr.20: 35472176-35472176 on Build GRCh38
rs764793910 Chr.20: 35472129-35472129 on Build GRCh38
rs761882466 Chr.20: 35472029-35472033 on Build GRCh38

View Full Product Details