Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41201162-41201423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP2S1
Entrez Gene ID
29785
Gene Name
cytochrome P450 family 2 subfamily S member 1
Gene Aliases
CYPIIS1
UniGene
Hs.98370
Gene Chromosome Location
Chr.19: 41193207-41207539 on Build GRCh38
Transcripts
NM_030622.7

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7126292 c.882G>A p.M294I Chr.19: 41201278-41201278 on Build GRCh38
6448381 c.967C>T p.H323Y Chr.19: 41201363-41201363 on Build GRCh38
996941 c.906T>C p.A302A Chr.19: 41201302-41201302 on Build GRCh38
3226680 c.964C>T p.P322S Chr.19: 41201360-41201360 on Build GRCh38
3534323 c.927C>T p.T309T Chr.19: 41201323-41201323 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762215456 Chr.19: 41201397-41201397 on Build GRCh38
rs545181272 Chr.19: 41201330-41201330 on Build GRCh38
rs958270412 Chr.19: 41201360-41201360 on Build GRCh38
rs756093596 Chr.19: 41201326-41201326 on Build GRCh38
rs749013053 Chr.19: 41201343-41201343 on Build GRCh38
rs774777865 Chr.19: 41201240-41201240 on Build GRCh38
rs777059573 Chr.19: 41201311-41201311 on Build GRCh38
rs749976098 Chr.19: 41201186-41201186 on Build GRCh38
rs151310483 Chr.19: 41201296-41201296 on Build GRCh38
rs769507751 Chr.19: 41201251-41201251 on Build GRCh38
rs140548760 Chr.19: 41201319-41201319 on Build GRCh38
rs373437581 Chr.19: 41201260-41201260 on Build GRCh38
rs748161742 Chr.19: 41201192-41201192 on Build GRCh38
rs745925929 Chr.19: 41201246-41201246 on Build GRCh38
rs754542013 Chr.19: 41201188-41201188 on Build GRCh38
rs767049063 Chr.19: 41201322-41201322 on Build GRCh38
rs757171848 Chr.19: 41201336-41201336 on Build GRCh38
rs749416592 Chr.19: 41201233-41201233 on Build GRCh38
rs780202651 Chr.19: 41201375-41201375 on Build GRCh38
rs145747863 Chr.19: 41201325-41201325 on Build GRCh38
rs137941427 Chr.19: 41201329-41201329 on Build GRCh38
rs191743223 Chr.19: 41201362-41201362 on Build GRCh38
rs777649108 Chr.19: 41201215-41201215 on Build GRCh38
rs950101301 Chr.19: 41201219-41201219 on Build GRCh38
rs755829362 Chr.19: 41201196-41201196 on Build GRCh38
rs772237721 Chr.19: 41201374-41201374 on Build GRCh38
rs927601225 Chr.19: 41201271-41201271 on Build GRCh38
rs747414886 Chr.19: 41201382-41201382 on Build GRCh38
rs768853500 Chr.19: 41201308-41201308 on Build GRCh38
rs139115184 Chr.19: 41201252-41201252 on Build GRCh38
rs143385223 Chr.19: 41201263-41201263 on Build GRCh38
rs766003336 Chr.19: 41201314-41201314 on Build GRCh38
rs762153062 Chr.19: 41201313-41201313 on Build GRCh38
rs779895334 Chr.19: 41201342-41201344 on Build GRCh38
rs879055848 Chr.19: 41201218-41201218 on Build GRCh38
rs761273716 Chr.19: 41201307-41201307 on Build GRCh38
rs372126413 Chr.19: 41201390-41201390 on Build GRCh38
rs756037606 Chr.19: 41201189-41201190 on Build GRCh38
rs1628289 Chr.19: 41201189-41201189 on Build GRCh38
rs899030207 Chr.19: 41201208-41201208 on Build GRCh38
rs751066695 Chr.19: 41201186-41201186 on Build GRCh38
rs771280888 Chr.19: 41201239-41201239 on Build GRCh38
rs769202831 Chr.19: 41201384-41201384 on Build GRCh38

View Full Product Details