Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2244267-2244522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ECI1
Entrez Gene ID
1632
Gene Name
enoyl-CoA delta isomerase 1
Gene Aliases
DCI
UniGene
Hs.403436
Gene Chromosome Location
Chr.16: 2239872-2251601 on Build GRCh38
Transcripts
NM_001178029.1 NM_001919.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5650069 c.439_441+2delAACGT p.? Chr.16: 2244404-2244408 on Build GRCh38
7417750 c.335A>T p.E112V Chr.16: 2244512-2244512 on Build GRCh38
4602755 c.382G>A p.V128I Chr.16: 2244465-2244465 on Build GRCh38
5642905 c.364G>A p.A122T Chr.16: 2244483-2244483 on Build GRCh38
7365819 c.421G>A p.V141M Chr.16: 2244426-2244426 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760423250 Chr.16: 2244445-2244445 on Build GRCh38
rs199619754 Chr.16: 2244365-2244365 on Build GRCh38
rs781561935 Chr.16: 2244414-2244414 on Build GRCh38
rs368708095 Chr.16: 2244372-2244372 on Build GRCh38
rs140753801 Chr.16: 2244439-2244439 on Build GRCh38
rs530685369 Chr.16: 2244310-2244310 on Build GRCh38
rs765276090 Chr.16: 2244449-2244449 on Build GRCh38
rs160555 Chr.16: 2244354-2244354 on Build GRCh38
rs764019052 Chr.16: 2244377-2244377 on Build GRCh38
rs150691746 Chr.16: 2244405-2244405 on Build GRCh38
rs550053066 Chr.16: 2244308-2244308 on Build GRCh38
rs772989313 Chr.16: 2244502-2244502 on Build GRCh38
rs779143999 Chr.16: 2244440-2244445 on Build GRCh38
rs909540211 Chr.16: 2244383-2244383 on Build GRCh38
rs201346166 Chr.16: 2244447-2244447 on Build GRCh38
rs541692981 Chr.16: 2244301-2244301 on Build GRCh38
rs765368036 Chr.16: 2244400-2244400 on Build GRCh38
rs746287914 Chr.16: 2244453-2244453 on Build GRCh38
rs532962324 Chr.16: 2244357-2244357 on Build GRCh38
rs144791724 Chr.16: 2244446-2244446 on Build GRCh38
rs745357478 Chr.16: 2244431-2244431 on Build GRCh38
rs774973333 Chr.16: 2244442-2244442 on Build GRCh38
rs373850326 Chr.16: 2244341-2244341 on Build GRCh38
rs753073208 Chr.16: 2244473-2244473 on Build GRCh38
rs757282014 Chr.16: 2244395-2244395 on Build GRCh38
rs538123254 Chr.16: 2244466-2244466 on Build GRCh38
rs1025716252 Chr.16: 2244421-2244421 on Build GRCh38
rs376657107 Chr.16: 2244492-2244492 on Build GRCh38
rs533971343 Chr.16: 2244484-2244484 on Build GRCh38
rs529477231 Chr.16: 2244407-2244407 on Build GRCh38
rs775271968 Chr.16: 2244370-2244370 on Build GRCh38
rs148548119 Chr.16: 2244493-2244493 on Build GRCh38
rs1020918518 Chr.16: 2244339-2244339 on Build GRCh38
rs372341953 Chr.16: 2244479-2244479 on Build GRCh38
rs544509062 Chr.16: 2244352-2244352 on Build GRCh38
rs748618022 Chr.16: 2244415-2244415 on Build GRCh38
rs200130130 Chr.16: 2244465-2244465 on Build GRCh38
rs946539530 Chr.16: 2244304-2244304 on Build GRCh38
rs775162611 Chr.16: 2244464-2244471 on Build GRCh38
rs753802399 Chr.16: 2244385-2244385 on Build GRCh38
rs756529820 Chr.16: 2244474-2244474 on Build GRCh38
rs778709886 Chr.16: 2244406-2244406 on Build GRCh38
rs377274938 Chr.16: 2244483-2244483 on Build GRCh38
rs769775669 Chr.16: 2244496-2244496 on Build GRCh38
rs771681200 Chr.16: 2244438-2244438 on Build GRCh38
rs773793866 Chr.16: 2244430-2244430 on Build GRCh38
rs200369056 Chr.16: 2244358-2244358 on Build GRCh38
rs771262093 Chr.16: 2244436-2244436 on Build GRCh38
rs771690070 Chr.16: 2244458-2244462 on Build GRCh38
rs142982644 Chr.16: 2244457-2244457 on Build GRCh38
rs958025460 Chr.16: 2244324-2244324 on Build GRCh38
rs994278121 Chr.16: 2244417-2244417 on Build GRCh38
rs771459648 Chr.16: 2244480-2244480 on Build GRCh38
rs761771407 Chr.16: 2244448-2244448 on Build GRCh38
rs762935665 Chr.16: 2244455-2244455 on Build GRCh38
rs778249875 Chr.16: 2244475-2244475 on Build GRCh38
rs915065835 Chr.16: 2244288-2244288 on Build GRCh38
rs572691056 Chr.16: 2244300-2244300 on Build GRCh38
rs750430412 Chr.16: 2244451-2244451 on Build GRCh38
rs770294190 Chr.16: 2244426-2244426 on Build GRCh38
rs1009568479 Chr.16: 2244327-2244327 on Build GRCh38
rs755277651 Chr.16: 2244412-2244412 on Build GRCh38

View Full Product Details