Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 186539386-186539625 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYGS
Entrez Gene ID
1427
Gene Name
crystallin gamma S
Gene Aliases
CRYG8, CTRCT20
UniGene
Hs.376209
Gene Chromosome Location
Chr.3: 186538443-186544378 on Build GRCh38
Transcripts
NM_017541.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3332288 c.44delA p.N15fs*20 Chr.3: 186539575-186539575 on Build GRCh38
3332287 c.75C>T p.C25C Chr.3: 186539544-186539544 on Build GRCh38
7362775 c.39C>A p.D13E Chr.3: 186539580-186539580 on Build GRCh38
7278263 c.59G>A p.R20H Chr.3: 186539560-186539560 on Build GRCh38
4115933 c.156G>A p.R52R Chr.3: 186539463-186539463 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769351806 Chr.3: 186539582-186539582 on Build GRCh38
rs375511065 Chr.3: 186539428-186539428 on Build GRCh38
rs928671942 Chr.3: 186539406-186539406 on Build GRCh38
rs777887949 Chr.3: 186539444-186539444 on Build GRCh38
rs750236806 Chr.3: 186539405-186539405 on Build GRCh38
rs550784657 Chr.3: 186539438-186539438 on Build GRCh38
rs772514521 Chr.3: 186539494-186539494 on Build GRCh38
rs535854737 Chr.3: 186539527-186539527 on Build GRCh38
rs774239734 Chr.3: 186539598-186539598 on Build GRCh38
rs143507827 Chr.3: 186539542-186539542 on Build GRCh38
rs767410546 Chr.3: 186539546-186539546 on Build GRCh38
rs750658278 Chr.3: 186539563-186539563 on Build GRCh38
rs772540045 Chr.3: 186539584-186539584 on Build GRCh38
rs144812537 Chr.3: 186539404-186539404 on Build GRCh38
rs779171765 Chr.3: 186539465-186539465 on Build GRCh38
rs761665528 Chr.3: 186539516-186539516 on Build GRCh38
rs143424000 Chr.3: 186539587-186539587 on Build GRCh38
rs757509772 Chr.3: 186539457-186539457 on Build GRCh38
rs569165861 Chr.3: 186539500-186539500 on Build GRCh38
rs759535446 Chr.3: 186539544-186539544 on Build GRCh38
rs148842396 Chr.3: 186539524-186539524 on Build GRCh38
rs752539198 Chr.3: 186539448-186539448 on Build GRCh38
rs753964791 Chr.3: 186539559-186539559 on Build GRCh38
rs761387196 Chr.3: 186539511-186539511 on Build GRCh38
rs746024951 Chr.3: 186539491-186539491 on Build GRCh38
rs776324135 Chr.3: 186539508-186539508 on Build GRCh38
rs867949981 Chr.3: 186539483-186539483 on Build GRCh38
rs200921733 Chr.3: 186539430-186539430 on Build GRCh38
rs747349708 Chr.3: 186539568-186539568 on Build GRCh38
rs113031838 Chr.3: 186539595-186539595 on Build GRCh38
rs147971015 Chr.3: 186539580-186539580 on Build GRCh38
rs369547458 Chr.3: 186539502-186539502 on Build GRCh38
rs757348269 Chr.3: 186539560-186539560 on Build GRCh38
rs776476771 Chr.3: 186539431-186539432 on Build GRCh38
rs762447393 Chr.3: 186539520-186539520 on Build GRCh38
rs1043437743 Chr.3: 186539555-186539555 on Build GRCh38
rs754108975 Chr.3: 186539537-186539538 on Build GRCh38
rs572718590 Chr.3: 186539565-186539565 on Build GRCh38
rs772949005 Chr.3: 186539513-186539513 on Build GRCh38
rs764793775 Chr.3: 186539512-186539512 on Build GRCh38
rs781770594 Chr.3: 186539579-186539579 on Build GRCh38
rs104893736 Chr.3: 186539566-186539566 on Build GRCh38
rs1035614113 Chr.3: 186539557-186539557 on Build GRCh38
rs769338745 Chr.3: 186539575-186539575 on Build GRCh38
rs1052019642 Chr.3: 186539422-186539422 on Build GRCh38
rs751519288 Chr.3: 186539431-186539431 on Build GRCh38
rs145500062 Chr.3: 186539501-186539501 on Build GRCh38
rs61743714 Chr.3: 186539564-186539564 on Build GRCh38
rs370574388 Chr.3: 186539561-186539561 on Build GRCh38
rs773186444 Chr.3: 186539572-186539600 on Build GRCh38
rs758237494 Chr.3: 186539416-186539416 on Build GRCh38

View Full Product Details