Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28997174-28997413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPB41
Entrez Gene ID
2035
Gene Name
erythrocyte membrane protein band 4.1
Gene Aliases
4.1R, EL1, HE
UniGene
Hs.175437
Gene Chromosome Location
Chr.1: 28887091-29120046 on Build GRCh38
Transcripts
NM_001166005.1 NM_001166006.1 NM_001166007.1 NM_004437.3 NM_203342.2 NM_203343.2 XM_005245753.1 XM_005245757.1 XM_005245760.1 XM_005245761.1 XM_005245763.1 XM_005245764.1 XM_005245765.1 XM_005245768.1 XM_005245769.1 XM_005245770.1 XM_005245772.4 XM_005245773.4 XM_005245774.1 XM_006710434.1 XM_006710439.1 XM_011540956.1 XM_011540957.1 XM_011540958.1 XM_011540959.1 XM_011540960.1 XM_011540961.1 XM_011540962.1 XM_011540963.2 XM_011540964.1 XM_011540965.2 XM_017000581.1 XM_017000582.1 XM_017000583.1 XM_017000584.1 XM_017000585.1 XM_017000586.1 XM_017000587.1 XM_017000588.1 XM_017000589.1 XM_017000590.1 XM_017000591.1 XM_017000592.1 XM_017000593.1 XM_017000594.1 XM_017000595.1 XM_017000596.1 XM_017000597.1 XM_017000598.1 XM_017000599.1 XM_017000600.1 XM_017000601.1 XM_017000602.1 XM_017000603.1 XM_017000604.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3386075 c.142G>C p.D48H Chr.1: 28997302-28997302 on Build GRCh38
6336285 c.159+8G>T p.? Chr.1: 28997327-28997327 on Build GRCh38
3719606 c.159+1G>A p.? Chr.1: 28997320-28997320 on Build GRCh38
7309726 c.684A>T p.K228N Chr.1: 28997217-28997217 on Build GRCh38
1341621 c.738G>A p.L246L Chr.1: 28997271-28997271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369527719 Chr.1: 28997250-28997250 on Build GRCh38
rs973969764 Chr.1: 28997326-28997326 on Build GRCh38
rs747566443 Chr.1: 28997294-28997294 on Build GRCh38
rs760586185 Chr.1: 28997235-28997235 on Build GRCh38
rs566540981 Chr.1: 28997308-28997308 on Build GRCh38
rs767443466 Chr.1: 28997205-28997205 on Build GRCh38
rs939263965 Chr.1: 28997359-28997363 on Build GRCh38
rs774386299 Chr.1: 28997338-28997338 on Build GRCh38
rs1024379477 Chr.1: 28997246-28997246 on Build GRCh38
rs929933368 Chr.1: 28997321-28997321 on Build GRCh38
rs552908580 Chr.1: 28997296-28997296 on Build GRCh38
rs138222761 Chr.1: 28997356-28997356 on Build GRCh38
rs768572526 Chr.1: 28997352-28997353 on Build GRCh38
rs770064316 Chr.1: 28997311-28997311 on Build GRCh38
rs773706226 Chr.1: 28997318-28997318 on Build GRCh38
rs752588508 Chr.1: 28997219-28997219 on Build GRCh38
rs756652002 Chr.1: 28997229-28997229 on Build GRCh38
rs780730982 Chr.1: 28997288-28997288 on Build GRCh38
rs751877537 Chr.1: 28997312-28997312 on Build GRCh38
rs146976856 Chr.1: 28997322-28997322 on Build GRCh38
rs756996105 Chr.1: 28997253-28997253 on Build GRCh38
rs920901853 Chr.1: 28997347-28997347 on Build GRCh38
rs141351853 Chr.1: 28997201-28997201 on Build GRCh38
rs373464494 Chr.1: 28997264-28997264 on Build GRCh38
rs781668873 Chr.1: 28997307-28997307 on Build GRCh38
rs763919964 Chr.1: 28997248-28997248 on Build GRCh38
rs762880453 Chr.1: 28997203-28997203 on Build GRCh38
rs769903319 Chr.1: 28997200-28997200 on Build GRCh38
rs749414197 Chr.1: 28997315-28997315 on Build GRCh38
rs764009856 Chr.1: 28997358-28997358 on Build GRCh38
rs757942511 Chr.1: 28997284-28997284 on Build GRCh38
rs894951783 Chr.1: 28997384-28997384 on Build GRCh38
rs377672093 Chr.1: 28997371-28997371 on Build GRCh38
rs749954369 Chr.1: 28997274-28997274 on Build GRCh38

View Full Product Details