Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 87610728-87610997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSPP
Entrez Gene ID
1834
Gene Name
dentin sialophosphoprotein
Gene Aliases
DFNA39, DGI1, DMP3, DPP, DSP
UniGene
Hs.678914
Gene Chromosome Location
Chr.4: 87608529-87616873 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6905492 c.29G>T p.W10L Chr.4: 87610937-87610937 on Build GRCh38
6900888 c.31G>A p.A11T Chr.4: 87610939-87610939 on Build GRCh38
6649679 c.44C>T p.A15V Chr.4: 87610952-87610952 on Build GRCh38
735398 c.29G>A p.W10* Chr.4: 87610937-87610937 on Build GRCh38
6284581 c.14C>T p.T5I Chr.4: 87610922-87610922 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754575103 Chr.4: 87610901-87610901 on Build GRCh38
rs897312874 Chr.4: 87610886-87610886 on Build GRCh38
rs121912989 Chr.4: 87610952-87610952 on Build GRCh38
rs946703693 Chr.4: 87610760-87610760 on Build GRCh38
rs750508334 Chr.4: 87610884-87610885 on Build GRCh38
rs557482020 Chr.4: 87610776-87610776 on Build GRCh38
rs778650282 Chr.4: 87610927-87610927 on Build GRCh38
rs566374136 Chr.4: 87610837-87610837 on Build GRCh38
rs759835461 Chr.4: 87610764-87610764 on Build GRCh38
rs746337396 Chr.4: 87610964-87610964 on Build GRCh38
rs769956177 Chr.4: 87610965-87610965 on Build GRCh38
rs892450520 Chr.4: 87610750-87610750 on Build GRCh38
rs941412720 Chr.4: 87610782-87610782 on Build GRCh38
rs747384492 Chr.4: 87610940-87610940 on Build GRCh38
rs28469148 Chr.4: 87610749-87610749 on Build GRCh38
rs1043799549 Chr.4: 87610771-87610771 on Build GRCh38
rs895379196 Chr.4: 87610944-87610944 on Build GRCh38
rs771383345 Chr.4: 87610943-87610943 on Build GRCh38
rs765748583 Chr.4: 87610869-87610869 on Build GRCh38
rs775427643 Chr.4: 87610966-87610966 on Build GRCh38
rs750314940 Chr.4: 87610876-87610876 on Build GRCh38
rs767443436 Chr.4: 87610862-87610862 on Build GRCh38
rs878886696 Chr.4: 87610950-87610950 on Build GRCh38
rs777010097 Chr.4: 87610762-87610762 on Build GRCh38
rs1037109925 Chr.4: 87610867-87610867 on Build GRCh38
rs1057049906 Chr.4: 87610899-87610899 on Build GRCh38
rs781578036 Chr.4: 87610894-87610894 on Build GRCh38
rs539358130 Chr.4: 87610761-87610761 on Build GRCh38
rs749040560 Chr.4: 87610898-87610898 on Build GRCh38
rs121912988 Chr.4: 87610924-87610924 on Build GRCh38
rs777298191 Chr.4: 87610963-87610963 on Build GRCh38
rs756985353 Chr.4: 87610859-87610859 on Build GRCh38
rs763290840 Chr.4: 87610968-87610968 on Build GRCh38
rs1003698155 Chr.4: 87610887-87610887 on Build GRCh38
rs121912986 Chr.4: 87610957-87610957 on Build GRCh38

View Full Product Details