Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49488203-49488436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPQ
Entrez Gene ID
55166
Gene Name
centromere protein Q
Gene Aliases
C6orf139, CENP-Q
UniGene
Hs.88663
Gene Chromosome Location
Chr.6: 49463327-49493107 on Build GRCh38
Transcripts
NM_018132.3 XM_005249205.1 XM_011514723.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6871293 c.495G>A p.M165I Chr.6: 49488369-49488369 on Build GRCh38
3697900 c.538A>C p.K180Q Chr.6: 49488412-49488412 on Build GRCh38
1200762 c.534C>A p.S178R Chr.6: 49488408-49488408 on Build GRCh38
5557717 c.490A>G p.K164E Chr.6: 49488364-49488364 on Build GRCh38
6626690 c.521G>T p.G174V Chr.6: 49488395-49488395 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993202707 Chr.6: 49488253-49488253 on Build GRCh38
rs547679783 Chr.6: 49488265-49488265 on Build GRCh38
rs761952956 Chr.6: 49488336-49488336 on Build GRCh38
rs758699241 Chr.6: 49488363-49488363 on Build GRCh38
rs551546106 Chr.6: 49488358-49488358 on Build GRCh38
rs987313313 Chr.6: 49488243-49488243 on Build GRCh38
rs1025907160 Chr.6: 49488256-49488256 on Build GRCh38
rs565871484 Chr.6: 49488289-49488289 on Build GRCh38
rs761202102 Chr.6: 49488328-49488329 on Build GRCh38
rs751863775 Chr.6: 49488372-49488372 on Build GRCh38
rs760734913 Chr.6: 49488303-49488303 on Build GRCh38
rs911043868 Chr.6: 49488255-49488255 on Build GRCh38
rs143879042 Chr.6: 49488276-49488276 on Build GRCh38
rs780396539 Chr.6: 49488364-49488364 on Build GRCh38
rs765587318 Chr.6: 49488342-49488342 on Build GRCh38
rs539218382 Chr.6: 49488295-49488295 on Build GRCh38
rs754018059 Chr.6: 49488320-49488320 on Build GRCh38
rs764108875 Chr.6: 49488307-49488307 on Build GRCh38
rs755530043 Chr.6: 49488393-49488393 on Build GRCh38
rs766835585 Chr.6: 49488329-49488329 on Build GRCh38
rs1038209728 Chr.6: 49488285-49488285 on Build GRCh38
rs773300924 Chr.6: 49488328-49488328 on Build GRCh38
rs960070396 Chr.6: 49488252-49488252 on Build GRCh38

View Full Product Details