Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63585120-63585385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF7
Entrez Gene ID
10238
Gene Name
DDB1 and CUL4 associated factor 7
Gene Aliases
AN11, HAN11, SWAN-1, WDR68
UniGene
Hs.410596
Gene Chromosome Location
Chr.17: 63550435-63594279 on Build GRCh38
Transcripts
NM_005828.4 NR_073585.1

SNP Detail

SNP ID Chromosome Location
rs759378485 Chr.17: 63585335-63585335 on Build GRCh38
rs759503157 Chr.17: 63585181-63585181 on Build GRCh38
rs755691700 Chr.17: 63585231-63585231 on Build GRCh38
rs572453308 Chr.17: 63585327-63585327 on Build GRCh38
rs763507354 Chr.17: 63585237-63585237 on Build GRCh38
rs563744514 Chr.17: 63585321-63585321 on Build GRCh38
rs767269687 Chr.17: 63585341-63585341 on Build GRCh38
rs775564315 Chr.17: 63585349-63585349 on Build GRCh38
rs78760999 Chr.17: 63585307-63585307 on Build GRCh38
rs765481363 Chr.17: 63585273-63585273 on Build GRCh38
rs774157140 Chr.17: 63585331-63585331 on Build GRCh38
rs768843071 Chr.17: 63585309-63585309 on Build GRCh38
rs2292422 Chr.17: 63585249-63585249 on Build GRCh38
rs200265994 Chr.17: 63585206-63585206 on Build GRCh38
rs746192971 Chr.17: 63585277-63585277 on Build GRCh38
rs544112424 Chr.17: 63585361-63585361 on Build GRCh38
rs925624922 Chr.17: 63585226-63585226 on Build GRCh38
rs774069397 Chr.17: 63585174-63585174 on Build GRCh38
rs752558835 Chr.17: 63585207-63585207 on Build GRCh38
rs766216566 Chr.17: 63585167-63585167 on Build GRCh38
rs540674406 Chr.17: 63585300-63585300 on Build GRCh38
rs781237623 Chr.17: 63585172-63585173 on Build GRCh38
rs35176162 Chr.17: 63585333-63585334 on Build GRCh38
rs375485650 Chr.17: 63585270-63585270 on Build GRCh38
rs1055853999 Chr.17: 63585342-63585342 on Build GRCh38
rs761934291 Chr.17: 63585166-63585166 on Build GRCh38
rs758995656 Chr.17: 63585281-63585281 on Build GRCh38

View Full Product Details