Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 92931279-92931530 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHGA
Entrez Gene ID
1113
Gene Name
chromogranin A
Gene Aliases
CGA
UniGene
Hs.150793
Gene Chromosome Location
Chr.14: 92922589-92935293 on Build GRCh38
Transcripts
NM_001275.3 NM_001301690.1 XM_011536370.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1201043 c.539C>T p.A180V Chr.14: 92931433-92931433 on Build GRCh38
4794965 c.508C>T p.P170S Chr.14: 92931402-92931402 on Build GRCh38
4384832 c.608G>C p.S203T Chr.14: 92931502-92931502 on Build GRCh38
4414236 c.436A>G p.T146A Chr.14: 92931330-92931330 on Build GRCh38
4880349 c.525G>C p.E175D Chr.14: 92931419-92931419 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765584956 Chr.14: 92931437-92931437 on Build GRCh38
rs144862632 Chr.14: 92931383-92931383 on Build GRCh38
rs779208725 Chr.14: 92931359-92931359 on Build GRCh38
rs758599091 Chr.14: 92931443-92931443 on Build GRCh38
rs1029637941 Chr.14: 92931441-92931441 on Build GRCh38
rs556109764 Chr.14: 92931392-92931392 on Build GRCh38
rs751670420 Chr.14: 92931458-92931458 on Build GRCh38
rs767848158 Chr.14: 92931323-92931323 on Build GRCh38
rs567354524 Chr.14: 92931335-92931335 on Build GRCh38
rs761219395 Chr.14: 92931419-92931419 on Build GRCh38
rs140821122 Chr.14: 92931309-92931309 on Build GRCh38
rs755398500 Chr.14: 92931428-92931428 on Build GRCh38
rs575865271 Chr.14: 92931396-92931396 on Build GRCh38
rs769762955 Chr.14: 92931387-92931387 on Build GRCh38
rs749018702 Chr.14: 92931475-92931475 on Build GRCh38
rs138912559 Chr.14: 92931337-92931337 on Build GRCh38
rs985572219 Chr.14: 92931449-92931449 on Build GRCh38
rs777878169 Chr.14: 92931445-92931445 on Build GRCh38
rs750925839 Chr.14: 92931324-92931324 on Build GRCh38
rs946677734 Chr.14: 92931400-92931400 on Build GRCh38
rs748107755 Chr.14: 92931374-92931374 on Build GRCh38
rs772725203 Chr.14: 92931507-92931507 on Build GRCh38
rs756187280 Chr.14: 92931414-92931422 on Build GRCh38
rs752949719 Chr.14: 92931442-92931442 on Build GRCh38
rs751677843 Chr.14: 92931413-92931414 on Build GRCh38
rs774060250 Chr.14: 92931410-92931410 on Build GRCh38
rs766510712 Chr.14: 92931414-92931419 on Build GRCh38
rs777775128 Chr.14: 92931381-92931381 on Build GRCh38
rs578186826 Chr.14: 92931498-92931498 on Build GRCh38
rs778618602 Chr.14: 92931497-92931497 on Build GRCh38
rs754195897 Chr.14: 92931422-92931422 on Build GRCh38
rs760274611 Chr.14: 92931510-92931510 on Build GRCh38
rs761402294 Chr.14: 92931312-92931312 on Build GRCh38
rs145227561 Chr.14: 92931414-92931416 on Build GRCh38
rs756000508 Chr.14: 92931462-92931462 on Build GRCh38
rs376742669 Chr.14: 92931311-92931311 on Build GRCh38
rs544894375 Chr.14: 92931418-92931418 on Build GRCh38
rs771649968 Chr.14: 92931502-92931502 on Build GRCh38
rs755202160 Chr.14: 92931358-92931358 on Build GRCh38
rs9658654 Chr.14: 92931420-92931420 on Build GRCh38
rs768439126 Chr.14: 92931492-92931492 on Build GRCh38
rs138696811 Chr.14: 92931389-92931389 on Build GRCh38
rs754281765 Chr.14: 92931351-92931351 on Build GRCh38
rs779983956 Chr.14: 92931464-92931464 on Build GRCh38
rs761441888 Chr.14: 92931416-92931416 on Build GRCh38
rs150148299 Chr.14: 92931310-92931310 on Build GRCh38
rs988620296 Chr.14: 92931349-92931349 on Build GRCh38
rs150244309 Chr.14: 92931336-92931336 on Build GRCh38
rs17847012 Chr.14: 92931329-92931329 on Build GRCh38

View Full Product Details