Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67373459-67373712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCF1
Entrez Gene ID
23529
Gene Name
cardiotrophin-like cytokine factor 1
Gene Aliases
BSF-3, BSF3, CISS2, CLC, NNT-1, NNT1, NR6
UniGene
Hs.502977
Gene Chromosome Location
Chr.11: 67364168-67374177 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

SNP Detail

SNP ID Chromosome Location
rs760653165 Chr.11: 67373531-67373531 on Build GRCh38
rs1054884915 Chr.11: 67373503-67373503 on Build GRCh38
rs887392779 Chr.11: 67373534-67373534 on Build GRCh38
rs1034927840 Chr.11: 67373591-67373591 on Build GRCh38
rs554428933 Chr.11: 67373600-67373600 on Build GRCh38
rs765530482 Chr.11: 67373554-67373554 on Build GRCh38
rs769668426 Chr.11: 67373492-67373492 on Build GRCh38
rs758862272 Chr.11: 67373571-67373571 on Build GRCh38
rs1021355319 Chr.11: 67373655-67373656 on Build GRCh38
rs779594800 Chr.11: 67373633-67373633 on Build GRCh38
rs1001972340 Chr.11: 67373527-67373527 on Build GRCh38
rs766488948 Chr.11: 67373532-67373532 on Build GRCh38
rs960581179 Chr.11: 67373593-67373593 on Build GRCh38
rs989513901 Chr.11: 67373645-67373645 on Build GRCh38
rs747599800 Chr.11: 67373589-67373589 on Build GRCh38
rs1028369328 Chr.11: 67373588-67373588 on Build GRCh38
rs778127677 Chr.11: 67373579-67373579 on Build GRCh38
rs755215558 Chr.11: 67373549-67373549 on Build GRCh38
rs112054504 Chr.11: 67373560-67373560 on Build GRCh38
rs762888312 Chr.11: 67373504-67373504 on Build GRCh38
rs754020360 Chr.11: 67373545-67373545 on Build GRCh38
rs775444420 Chr.11: 67373493-67373493 on Build GRCh38
rs763939968 Chr.11: 67373506-67373506 on Build GRCh38
rs980552478 Chr.11: 67373497-67373497 on Build GRCh38
rs765298833 Chr.11: 67373678-67373678 on Build GRCh38
rs996928744 Chr.11: 67373578-67373578 on Build GRCh38
rs774407498 Chr.11: 67373511-67373511 on Build GRCh38

View Full Product Details