Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122208478-122208705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIABLO
Entrez Gene ID
56616
Gene Name
diablo IAP-binding mitochondrial protein
Gene Aliases
DFNA64, SMAC
UniGene
Hs.169611
Gene Chromosome Location
Chr.12: 122207662-122227534 on Build GRCh38
Transcripts
NM_001278302.1 NM_001278303.1 NM_001278304.1 NM_001278342.1 NM_019887.5 NM_138930.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4502112 c.604C>T p.R202W Chr.12: 122208497-122208497 on Build GRCh38
936186 c.525C>T p.G175G Chr.12: 122208576-122208576 on Build GRCh38
117030 c.545C>A p.T182N Chr.12: 122208556-122208556 on Build GRCh38
369882 c.546C>T p.T182T Chr.12: 122208555-122208555 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372445889 Chr.12: 122208534-122208534 on Build GRCh38
rs781561232 Chr.12: 122208616-122208616 on Build GRCh38
rs377015837 Chr.12: 122208592-122208592 on Build GRCh38
rs760073847 Chr.12: 122208556-122208556 on Build GRCh38
rs200309542 Chr.12: 122208623-122208623 on Build GRCh38
rs191255872 Chr.12: 122208560-122208560 on Build GRCh38
rs752646534 Chr.12: 122208684-122208684 on Build GRCh38
rs772377484 Chr.12: 122208521-122208521 on Build GRCh38
rs746221208 Chr.12: 122208617-122208617 on Build GRCh38
rs750056920 Chr.12: 122208566-122208566 on Build GRCh38
rs764647908 Chr.12: 122208508-122208508 on Build GRCh38
rs749218903 Chr.12: 122208498-122208498 on Build GRCh38
rs759269464 Chr.12: 122208671-122208671 on Build GRCh38
rs865798313 Chr.12: 122208541-122208541 on Build GRCh38
rs748451619 Chr.12: 122208641-122208641 on Build GRCh38
rs745899792 Chr.12: 122208588-122208589 on Build GRCh38
rs763501543 Chr.12: 122208621-122208621 on Build GRCh38
rs894160258 Chr.12: 122208632-122208632 on Build GRCh38
rs371642727 Chr.12: 122208603-122208603 on Build GRCh38
rs535135642 Chr.12: 122208597-122208597 on Build GRCh38
rs748255110 Chr.12: 122208516-122208516 on Build GRCh38
rs747280328 Chr.12: 122208537-122208537 on Build GRCh38
rs1004131039 Chr.12: 122208673-122208673 on Build GRCh38
rs368429160 Chr.12: 122208507-122208507 on Build GRCh38
rs867016501 Chr.12: 122208513-122208513 on Build GRCh38
rs145271890 Chr.12: 122208639-122208639 on Build GRCh38
rs909700188 Chr.12: 122208501-122208501 on Build GRCh38
rs575550434 Chr.12: 122208572-122208572 on Build GRCh38
rs973478698 Chr.12: 122208565-122208565 on Build GRCh38
rs199738234 Chr.12: 122208548-122208548 on Build GRCh38
rs1025398339 Chr.12: 122208663-122208663 on Build GRCh38
rs767109973 Chr.12: 122208562-122208562 on Build GRCh38
rs748187773 Chr.12: 122208599-122208599 on Build GRCh38
rs375537524 Chr.12: 122208497-122208497 on Build GRCh38
rs201498504 Chr.12: 122208612-122208612 on Build GRCh38
rs995176953 Chr.12: 122208645-122208645 on Build GRCh38
rs375069899 Chr.12: 122208618-122208618 on Build GRCh38
rs770029584 Chr.12: 122208604-122208604 on Build GRCh38
rs368077506 Chr.12: 122208606-122208606 on Build GRCh38
rs914018547 Chr.12: 122208605-122208605 on Build GRCh38
rs369135778 Chr.12: 122208622-122208622 on Build GRCh38
rs773781886 Chr.12: 122208558-122208558 on Build GRCh38
rs1025291135 Chr.12: 122208682-122208682 on Build GRCh38
rs1052603444 Chr.12: 122208598-122208598 on Build GRCh38
rs753638530 Chr.12: 122208576-122208576 on Build GRCh38
rs753604950 Chr.12: 122208667-122208667 on Build GRCh38
rs372001893 Chr.12: 122208611-122208611 on Build GRCh38
rs779040136 Chr.12: 122208509-122208509 on Build GRCh38
rs765169855 Chr.12: 122208676-122208676 on Build GRCh38
rs1023960685 Chr.12: 122208657-122208657 on Build GRCh38
rs766195715 Chr.12: 122208575-122208575 on Build GRCh38
rs754889194 Chr.12: 122208589-122208589 on Build GRCh38
rs149387774 Chr.12: 122208555-122208555 on Build GRCh38
rs781358968 Chr.12: 122208554-122208554 on Build GRCh38

View Full Product Details