Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 104032126-104032399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL17A1
Entrez Gene ID
1308
Gene Name
collagen type XVII alpha 1
Gene Aliases
BA16H23.2, BP180, BPA-2, BPAG2, ERED, LAD-1
UniGene
Hs.117938
Gene Chromosome Location
Chr.10: 104031288-104085880 on Build GRCh38
Transcripts
NM_000494.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7163480 c.4485C>G p.V1495V Chr.10: 104032244-104032244 on Build GRCh38
5497987 c.4490C>T p.P1497L Chr.10: 104032239-104032239 on Build GRCh38
1201806 c.4475G>T p.S1492I Chr.10: 104032254-104032254 on Build GRCh38
6128093 c.4459C>A p.R1487R Chr.10: 104032270-104032270 on Build GRCh38
310225 c.4439G>A p.G1480D Chr.10: 104032290-104032290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12253626 Chr.10: 104032161-104032161 on Build GRCh38
rs369854778 Chr.10: 104032239-104032239 on Build GRCh38
rs761215469 Chr.10: 104032290-104032290 on Build GRCh38
rs756881229 Chr.10: 104032254-104032256 on Build GRCh38
rs781259999 Chr.10: 104032247-104032247 on Build GRCh38
rs769214913 Chr.10: 104032325-104032326 on Build GRCh38
rs780529528 Chr.10: 104032260-104032262 on Build GRCh38
rs805688 Chr.10: 104032319-104032319 on Build GRCh38
rs998546941 Chr.10: 104032363-104032363 on Build GRCh38
rs373388207 Chr.10: 104032271-104032271 on Build GRCh38
rs805687 Chr.10: 104032171-104032171 on Build GRCh38
rs777427203 Chr.10: 104032238-104032238 on Build GRCh38
rs750067078 Chr.10: 104032216-104032216 on Build GRCh38
rs766881854 Chr.10: 104032306-104032306 on Build GRCh38
rs1052870972 Chr.10: 104032351-104032351 on Build GRCh38
rs763164002 Chr.10: 104032256-104032256 on Build GRCh38
rs745763900 Chr.10: 104032251-104032251 on Build GRCh38
rs144719079 Chr.10: 104032263-104032263 on Build GRCh38
rs866213117 Chr.10: 104032368-104032368 on Build GRCh38
rs777629337 Chr.10: 104032323-104032323 on Build GRCh38
rs751170106 Chr.10: 104032324-104032324 on Build GRCh38
rs368513242 Chr.10: 104032226-104032226 on Build GRCh38
rs148509618 Chr.10: 104032269-104032269 on Build GRCh38
rs752421627 Chr.10: 104032230-104032230 on Build GRCh38
rs536571134 Chr.10: 104032232-104032232 on Build GRCh38
rs370488594 Chr.10: 104032327-104032327 on Build GRCh38
rs749613805 Chr.10: 104032263-104032265 on Build GRCh38
rs376525143 Chr.10: 104032307-104032307 on Build GRCh38
rs764186232 Chr.10: 104032192-104032192 on Build GRCh38
rs769597571 Chr.10: 104032253-104032253 on Build GRCh38
rs770337635 Chr.10: 104032244-104032244 on Build GRCh38
rs375121106 Chr.10: 104032194-104032194 on Build GRCh38
rs149754696 Chr.10: 104032227-104032227 on Build GRCh38
rs768127417 Chr.10: 104032209-104032209 on Build GRCh38
rs548548309 Chr.10: 104032255-104032255 on Build GRCh38
rs758119165 Chr.10: 104032317-104032317 on Build GRCh38
rs886046681 Chr.10: 104032178-104032178 on Build GRCh38
rs977419026 Chr.10: 104032348-104032348 on Build GRCh38
rs768510987 Chr.10: 104032356-104032356 on Build GRCh38
rs745846158 Chr.10: 104032338-104032338 on Build GRCh38
rs972823199 Chr.10: 104032208-104032208 on Build GRCh38
rs767463193 Chr.10: 104032282-104032282 on Build GRCh38
rs201704631 Chr.10: 104032199-104032199 on Build GRCh38
rs888118253 Chr.10: 104032178-104032178 on Build GRCh38
rs1015150064 Chr.10: 104032196-104032196 on Build GRCh38

View Full Product Details