Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37863159-37863406 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3L
Entrez Gene ID
51386
Gene Name
eukaryotic translation initiation factor 3 subunit L
Gene Aliases
EIF3EIP, EIF3S11, EIF3S6IP, HSPC021, HSPC025, MSTP005
UniGene
Hs.446852
Gene Chromosome Location
Chr.22: 37848774-37888782 on Build GRCh38
Transcripts
NM_001242923.1 NM_016091.3 XM_005261625.1 XM_006724260.3 XM_017028814.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652879 c.540C>T p.N180N Chr.22: 37863306-37863306 on Build GRCh38
4680626 c.510C>T p.A170A Chr.22: 37863276-37863276 on Build GRCh38
6652881 c.579G>T p.Q193H Chr.22: 37863345-37863345 on Build GRCh38
6162295 c.524C>G p.P175R Chr.22: 37863290-37863290 on Build GRCh38
4582395 c.690C>T p.I230I Chr.22: 37863327-37863327 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019247254 Chr.22: 37863214-37863214 on Build GRCh38
rs768219088 Chr.22: 37863286-37863286 on Build GRCh38
rs751540875 Chr.22: 37863308-37863308 on Build GRCh38
rs980171268 Chr.22: 37863256-37863256 on Build GRCh38
rs781320528 Chr.22: 37863354-37863359 on Build GRCh38
rs369186163 Chr.22: 37863384-37863384 on Build GRCh38
rs754119383 Chr.22: 37863352-37863352 on Build GRCh38
rs762385956 Chr.22: 37863282-37863282 on Build GRCh38
rs945235168 Chr.22: 37863339-37863339 on Build GRCh38
rs960311047 Chr.22: 37863318-37863318 on Build GRCh38
rs935992358 Chr.22: 37863276-37863276 on Build GRCh38
rs987631857 Chr.22: 37863209-37863209 on Build GRCh38
rs766895925 Chr.22: 37863336-37863336 on Build GRCh38
rs780827504 Chr.22: 37863244-37863244 on Build GRCh38
rs754708343 Chr.22: 37863312-37863312 on Build GRCh38
rs758385893 Chr.22: 37863224-37863224 on Build GRCh38
rs151317788 Chr.22: 37863327-37863327 on Build GRCh38
rs769328106 Chr.22: 37863255-37863255 on Build GRCh38
rs372075058 Chr.22: 37863236-37863236 on Build GRCh38
rs113036185 Chr.22: 37863346-37863346 on Build GRCh38
rs950957124 Chr.22: 37863198-37863198 on Build GRCh38
rs372316591 Chr.22: 37863354-37863354 on Build GRCh38
rs375153596 Chr.22: 37863309-37863309 on Build GRCh38
rs777701016 Chr.22: 37863230-37863230 on Build GRCh38
rs779963254 Chr.22: 37863277-37863277 on Build GRCh38
rs746880813 Chr.22: 37863235-37863235 on Build GRCh38
rs925955795 Chr.22: 37863268-37863268 on Build GRCh38
rs111677695 Chr.22: 37863271-37863271 on Build GRCh38
rs964940448 Chr.22: 37863215-37863215 on Build GRCh38
rs1046210904 Chr.22: 37863367-37863367 on Build GRCh38
rs752904926 Chr.22: 37863379-37863379 on Build GRCh38
rs989303246 Chr.22: 37863322-37863322 on Build GRCh38
rs767027126 Chr.22: 37863212-37863212 on Build GRCh38
rs777789278 Chr.22: 37863382-37863382 on Build GRCh38
rs376912297 Chr.22: 37863355-37863355 on Build GRCh38
rs765363056 Chr.22: 37863359-37863359 on Build GRCh38
rs745576237 Chr.22: 37863249-37863249 on Build GRCh38

View Full Product Details