Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 10187434-10187693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN4
Entrez Gene ID
1183
Gene Name
chloride voltage-gated channel 4
Gene Aliases
CLC4, ClC-4, ClC-4A
UniGene
Hs.495674
Gene Chromosome Location
Chr.X: 10156945-10237660 on Build GRCh38
Transcripts
NM_001256944.1 NM_001830.3

SNP Detail

SNP ID Chromosome Location
rs770381670 Chr.X: 10187504-10187504 on Build GRCh38
rs776088449 Chr.X: 10187463-10187463 on Build GRCh38
rs769217062 Chr.X: 10187661-10187661 on Build GRCh38
rs202245864 Chr.X: 10187511-10187511 on Build GRCh38
rs764804263 Chr.X: 10187576-10187576 on Build GRCh38
rs773821059 Chr.X: 10187506-10187506 on Build GRCh38
rs373441034 Chr.X: 10187472-10187472 on Build GRCh38
rs781439502 Chr.X: 10187623-10187623 on Build GRCh38
rs745988994 Chr.X: 10187466-10187466 on Build GRCh38
rs768904980 Chr.X: 10187481-10187481 on Build GRCh38
rs777737862 Chr.X: 10187601-10187601 on Build GRCh38
rs147134396 Chr.X: 10187580-10187580 on Build GRCh38
rs762246952 Chr.X: 10187494-10187494 on Build GRCh38
rs767240677 Chr.X: 10187532-10187532 on Build GRCh38
rs368823929 Chr.X: 10187471-10187471 on Build GRCh38
rs763491779 Chr.X: 10187642-10187642 on Build GRCh38
rs765986707 Chr.X: 10187519-10187519 on Build GRCh38
rs370726692 Chr.X: 10187641-10187641 on Build GRCh38
rs763425301 Chr.X: 10187632-10187632 on Build GRCh38
rs192492405 Chr.X: 10187618-10187618 on Build GRCh38
rs762498580 Chr.X: 10187660-10187660 on Build GRCh38
rs1000813985 Chr.X: 10187665-10187665 on Build GRCh38
rs140344775 Chr.X: 10187604-10187604 on Build GRCh38
rs766614915 Chr.X: 10187499-10187499 on Build GRCh38
rs781599689 Chr.X: 10187535-10187535 on Build GRCh38
rs200370235 Chr.X: 10187619-10187619 on Build GRCh38
rs751180181 Chr.X: 10187520-10187520 on Build GRCh38
rs776994137 Chr.X: 10187492-10187492 on Build GRCh38
rs919124115 Chr.X: 10187593-10187593 on Build GRCh38
rs759174182 Chr.X: 10187530-10187530 on Build GRCh38
rs188033970 Chr.X: 10187473-10187473 on Build GRCh38
rs373242158 Chr.X: 10187613-10187613 on Build GRCh38
rs374769266 Chr.X: 10187465-10187465 on Build GRCh38
rs770257143 Chr.X: 10187634-10187634 on Build GRCh38

View Full Product Details