Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 373446-373707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B4GALNT4
Entrez Gene ID
338707
Gene Name
beta-1,4-N-acetyl-galactosaminyltransferase 4
Gene Aliases
-
UniGene
Hs.148074
Gene Chromosome Location
Chr.11: 369773-382117 on Build GRCh38
Transcripts
NM_178537.4 XM_017017654.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6584914 c.704+3G>C p.? Chr.11: 373519-373519 on Build GRCh38
4441923 c.701G>T p.R234M Chr.11: 373513-373513 on Build GRCh38
5441179 c.699C>T p.P233P Chr.11: 373511-373511 on Build GRCh38
7414817 c.674A>G p.K225R Chr.11: 373486-373486 on Build GRCh38
6496479 c.703C>A p.R235R Chr.11: 373515-373515 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908563783 Chr.11: 373608-373609 on Build GRCh38
rs761536331 Chr.11: 373553-373553 on Build GRCh38
rs886617605 Chr.11: 373528-373528 on Build GRCh38
rs73400534 Chr.11: 373661-373661 on Build GRCh38
rs531747294 Chr.11: 373623-373623 on Build GRCh38
rs774886142 Chr.11: 373552-373552 on Build GRCh38
rs780203515 Chr.11: 373564-373564 on Build GRCh38
rs781387302 Chr.11: 373674-373674 on Build GRCh38
rs114861429 Chr.11: 373542-373542 on Build GRCh38
rs879403625 Chr.11: 373625-373625 on Build GRCh38
rs368683615 Chr.11: 373558-373558 on Build GRCh38
rs367880649 Chr.11: 373541-373541 on Build GRCh38
rs577592474 Chr.11: 373610-373610 on Build GRCh38
rs756990547 Chr.11: 373468-373468 on Build GRCh38
rs770326089 Chr.11: 373530-373530 on Build GRCh38
rs1011299258 Chr.11: 373544-373544 on Build GRCh38
rs151250072 Chr.11: 373469-373469 on Build GRCh38
rs142833372 Chr.11: 373615-373615 on Build GRCh38
rs369181766 Chr.11: 373628-373628 on Build GRCh38
rs563210112 Chr.11: 373616-373616 on Build GRCh38
rs199586034 Chr.11: 373516-373516 on Build GRCh38
rs1012454807 Chr.11: 373675-373675 on Build GRCh38
rs778705905 Chr.11: 373513-373513 on Build GRCh38
rs138927306 Chr.11: 373605-373605 on Build GRCh38
rs745699589 Chr.11: 373554-373554 on Build GRCh38
rs1025146545 Chr.11: 373682-373682 on Build GRCh38
rs556059016 Chr.11: 373680-373680 on Build GRCh38
rs73400533 Chr.11: 373499-373499 on Build GRCh38
rs1047487214 Chr.11: 373634-373634 on Build GRCh38
rs970661703 Chr.11: 373573-373573 on Build GRCh38
rs765930777 Chr.11: 373565-373565 on Build GRCh38
rs569833363 Chr.11: 373470-373470 on Build GRCh38
rs756066552 Chr.11: 373476-373476 on Build GRCh38
rs901500187 Chr.11: 373511-373511 on Build GRCh38
rs1024969519 Chr.11: 373572-373572 on Build GRCh38
rs76656827 Chr.11: 373627-373627 on Build GRCh38
rs775793596 Chr.11: 373537-373537 on Build GRCh38
rs934529513 Chr.11: 373602-373602 on Build GRCh38
rs921340107 Chr.11: 373624-373624 on Build GRCh38
rs138345753 Chr.11: 373515-373515 on Build GRCh38
rs754713999 Chr.11: 373509-373509 on Build GRCh38
rs753674304 Chr.11: 373568-373568 on Build GRCh38
rs923990556 Chr.11: 373598-373598 on Build GRCh38
rs763336894 Chr.11: 373581-373586 on Build GRCh38
rs768866573 Chr.11: 373652-373652 on Build GRCh38
rs745989863 Chr.11: 373526-373526 on Build GRCh38
rs115079388 Chr.11: 373492-373492 on Build GRCh38
rs374254969 Chr.11: 373557-373557 on Build GRCh38
rs978153658 Chr.11: 373591-373591 on Build GRCh38
rs776748101 Chr.11: 373525-373525 on Build GRCh38
rs1031305402 Chr.11: 373536-373536 on Build GRCh38
rs76847575 Chr.11: 373664-373664 on Build GRCh38

View Full Product Details