Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 1997221-1997494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD2
Entrez Gene ID
55643
Gene Name
BTB domain containing 2
Gene Aliases
-
UniGene
Hs.465543
Gene Chromosome Location
Chr.19: 1985438-2015703 on Build GRCh38
Transcripts
NM_017797.3 XM_005259593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5747958 c.467C>T p.T156I Chr.19: 1997404-1997404 on Build GRCh38
4913576 c.445A>G p.M149V Chr.19: 1997426-1997426 on Build GRCh38
993438 c.443C>T p.A148V Chr.19: 1997428-1997428 on Build GRCh38
5580364 c.510C>T p.A170A Chr.19: 1997361-1997361 on Build GRCh38
6613355 c.453C>T p.N151N Chr.19: 1997418-1997418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764500555 Chr.19: 1997351-1997351 on Build GRCh38
rs567664956 Chr.19: 1997283-1997283 on Build GRCh38
rs754684263 Chr.19: 1997428-1997428 on Build GRCh38
rs933251944 Chr.19: 1997383-1997383 on Build GRCh38
rs748116433 Chr.19: 1997299-1997299 on Build GRCh38
rs777810607 Chr.19: 1997450-1997450 on Build GRCh38
rs918124582 Chr.19: 1997318-1997318 on Build GRCh38
rs771243506 Chr.19: 1997331-1997331 on Build GRCh38
rs762938064 Chr.19: 1997322-1997322 on Build GRCh38
rs766008900 Chr.19: 1997418-1997418 on Build GRCh38
rs536055395 Chr.19: 1997273-1997273 on Build GRCh38
rs760930391 Chr.19: 1997402-1997402 on Build GRCh38
rs746114134 Chr.19: 1997388-1997388 on Build GRCh38
rs775896833 Chr.19: 1997395-1997395 on Build GRCh38
rs773347928 Chr.19: 1997319-1997319 on Build GRCh38
rs752901269 Chr.19: 1997353-1997353 on Build GRCh38
rs747833020 Chr.19: 1997366-1997366 on Build GRCh38
rs141098024 Chr.19: 1997459-1997459 on Build GRCh38
rs141725644 Chr.19: 1997471-1997471 on Build GRCh38
rs139658440 Chr.19: 1997282-1997282 on Build GRCh38
rs200650426 Chr.19: 1997414-1997414 on Build GRCh38
rs143677178 Chr.19: 1997355-1997355 on Build GRCh38
rs747440617 Chr.19: 1997470-1997470 on Build GRCh38
rs759938169 Chr.19: 1997333-1997333 on Build GRCh38
rs769561691 Chr.19: 1997301-1997301 on Build GRCh38
rs761245147 Chr.19: 1997347-1997347 on Build GRCh38
rs968069025 Chr.19: 1997280-1997280 on Build GRCh38
rs891593686 Chr.19: 1997265-1997265 on Build GRCh38
rs113296918 Chr.19: 1997384-1997384 on Build GRCh38
rs151104877 Chr.19: 1997442-1997442 on Build GRCh38
rs772334258 Chr.19: 1997460-1997460 on Build GRCh38
rs781526303 Chr.19: 1997281-1997281 on Build GRCh38
rs776972502 Chr.19: 1997416-1997416 on Build GRCh38
rs775771939 Chr.19: 1997345-1997345 on Build GRCh38
rs750003411 Chr.19: 1997352-1997352 on Build GRCh38
rs577597760 Chr.19: 1997367-1997367 on Build GRCh38
rs767276168 Chr.19: 1997438-1997438 on Build GRCh38
rs762238863 Chr.19: 1997472-1997472 on Build GRCh38
rs1030170097 Chr.19: 1997262-1997262 on Build GRCh38
rs912601773 Chr.19: 1997292-1997292 on Build GRCh38
rs1009693931 Chr.19: 1997270-1997270 on Build GRCh38
rs749593700 Chr.19: 1997379-1997379 on Build GRCh38
rs762340181 Chr.19: 1997417-1997417 on Build GRCh38
rs1025644428 Chr.19: 1997277-1997277 on Build GRCh38
rs575845390 Chr.19: 1997421-1997421 on Build GRCh38
rs186972171 Chr.19: 1997394-1997394 on Build GRCh38
rs113648977 Chr.19: 1997376-1997376 on Build GRCh38
rs144229360 Chr.19: 1997439-1997439 on Build GRCh38
rs766174396 Chr.19: 1997354-1997354 on Build GRCh38
rs200511904 Chr.19: 1997332-1997332 on Build GRCh38
rs995649376 Chr.19: 1997259-1997259 on Build GRCh38
rs749219337 Chr.19: 1997451-1997451 on Build GRCh38
rs1001323700 Chr.19: 1997445-1997445 on Build GRCh38
rs767903520 Chr.19: 1997338-1997338 on Build GRCh38
rs775550147 Chr.19: 1997466-1997466 on Build GRCh38

View Full Product Details