Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43364207-43364472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELOVL1
Entrez Gene ID
64834
Gene Name
ELOVL fatty acid elongase 1
Gene Aliases
CGI-88, Ssc1
UniGene
Hs.25597
Gene Chromosome Location
Chr.1: 43363397-43368074 on Build GRCh38
Transcripts
NM_001256399.1 NM_001256401.1 NM_001256402.1 NM_022821.3 NR_046117.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680819 c.515C>T p.S172F Chr.1: 43364427-43364427 on Build GRCh38
6036888 c.618+8G>A p.? Chr.1: 43364316-43364316 on Build GRCh38
4566864 c.498_499CC>TT p.H167Y Chr.1: 43364443-43364444 on Build GRCh38
269093 c.601A>G p.M201V Chr.1: 43364341-43364341 on Build GRCh38
909761 c.602T>C p.M201T Chr.1: 43364340-43364340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748707708 Chr.1: 43364422-43364422 on Build GRCh38
rs766504794 Chr.1: 43364258-43364258 on Build GRCh38
rs367596149 Chr.1: 43364298-43364298 on Build GRCh38
rs762710876 Chr.1: 43364357-43364357 on Build GRCh38
rs779836848 Chr.1: 43364283-43364283 on Build GRCh38
rs939999689 Chr.1: 43364349-43364349 on Build GRCh38
rs35409714 Chr.1: 43364439-43364440 on Build GRCh38
rs757333738 Chr.1: 43364275-43364275 on Build GRCh38
rs751690290 Chr.1: 43364329-43364329 on Build GRCh38
rs908498464 Chr.1: 43364318-43364318 on Build GRCh38
rs746756197 Chr.1: 43364285-43364285 on Build GRCh38
rs772533745 Chr.1: 43364341-43364341 on Build GRCh38
rs747672101 Chr.1: 43364316-43364316 on Build GRCh38
rs746856215 Chr.1: 43364450-43364450 on Build GRCh38
rs758642204 Chr.1: 43364429-43364429 on Build GRCh38
rs780209491 Chr.1: 43364437-43364437 on Build GRCh38
rs772213579 Chr.1: 43364280-43364280 on Build GRCh38
rs754354091 Chr.1: 43364402-43364402 on Build GRCh38
rs753901619 Chr.1: 43364274-43364274 on Build GRCh38
rs113928928 Chr.1: 43364426-43364426 on Build GRCh38
rs371472640 Chr.1: 43364378-43364378 on Build GRCh38
rs746097010 Chr.1: 43364279-43364279 on Build GRCh38
rs571173066 Chr.1: 43364418-43364418 on Build GRCh38
rs1029930312 Chr.1: 43364413-43364413 on Build GRCh38
rs372744275 Chr.1: 43364254-43364254 on Build GRCh38
rs932831697 Chr.1: 43364420-43364420 on Build GRCh38
rs45600435 Chr.1: 43364295-43364295 on Build GRCh38
rs11558088 Chr.1: 43364379-43364379 on Build GRCh38
rs1036945812 Chr.1: 43364371-43364371 on Build GRCh38
rs761086443 Chr.1: 43364388-43364388 on Build GRCh38
rs766083833 Chr.1: 43364375-43364375 on Build GRCh38
rs977033100 Chr.1: 43364365-43364365 on Build GRCh38
rs150570611 Chr.1: 43364396-43364396 on Build GRCh38
rs769343809 Chr.1: 43364322-43364322 on Build GRCh38
rs757345481 Chr.1: 43364408-43364408 on Build GRCh38
rs200219758 Chr.1: 43364423-43364423 on Build GRCh38
rs779059720 Chr.1: 43364276-43364276 on Build GRCh38

View Full Product Details