Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87884468-87884729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC497256
Entrez Gene ID
497256
Gene Name
uncharacterized LOC497256
Gene Aliases
-
UniGene
Hs.584108
Gene Chromosome Location
Chr.9: 87844517-87939244 on Build GRCh38
Transcripts
NM_178828.4

Gene Symbol
SPATA31E1
Entrez Gene ID
286234
Gene Name
SPATA31 subfamily E member 1
Gene Aliases
C9orf79, FAM75E1
UniGene
Hs.130672
Gene Chromosome Location
Chr.9: 87882857-87888899 on Build GRCh38
Transcripts
NM_178828.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4854856 c.400C>T p.R134W Chr.9: 87884626-87884626 on Build GRCh38
341561 c.405C>A p.D135E Chr.9: 87884631-87884631 on Build GRCh38
3659343 c.375C>T p.I125I Chr.9: 87884601-87884601 on Build GRCh38
6788276 c.419T>C p.L140P Chr.9: 87884645-87884645 on Build GRCh38
3908708 c.380T>C p.L127P Chr.9: 87884606-87884606 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs555448052 Chr.9: 87884565-87884565 on Build GRCh38
rs752754642 Chr.9: 87884638-87884638 on Build GRCh38
rs769846206 Chr.9: 87884549-87884549 on Build GRCh38
rs546069187 Chr.9: 87884709-87884709 on Build GRCh38
rs763594758 Chr.9: 87884567-87884567 on Build GRCh38
rs756479213 Chr.9: 87884511-87884511 on Build GRCh38
rs985513939 Chr.9: 87884689-87884689 on Build GRCh38
rs34056147 Chr.9: 87884497-87884497 on Build GRCh38
rs373979437 Chr.9: 87884560-87884560 on Build GRCh38
rs769511302 Chr.9: 87884692-87884692 on Build GRCh38
rs554774274 Chr.9: 87884489-87884489 on Build GRCh38
rs770318836 Chr.9: 87884598-87884598 on Build GRCh38
rs772065495 Chr.9: 87884561-87884561 on Build GRCh38
rs73653971 Chr.9: 87884604-87884604 on Build GRCh38
rs756385926 Chr.9: 87884647-87884647 on Build GRCh38
rs952190487 Chr.9: 87884635-87884635 on Build GRCh38
rs774197042 Chr.9: 87884554-87884554 on Build GRCh38
rs754356164 Chr.9: 87884614-87884614 on Build GRCh38
rs776327099 Chr.9: 87884571-87884571 on Build GRCh38
rs777806175 Chr.9: 87884661-87884661 on Build GRCh38
rs761468855 Chr.9: 87884572-87884572 on Build GRCh38
rs776123880 Chr.9: 87884678-87884678 on Build GRCh38
rs1043573031 Chr.9: 87884568-87884568 on Build GRCh38
rs748367549 Chr.9: 87884547-87884547 on Build GRCh38
rs775270425 Chr.9: 87884564-87884564 on Build GRCh38
rs763439303 Chr.9: 87884697-87884697 on Build GRCh38
rs771429495 Chr.9: 87884668-87884669 on Build GRCh38
rs368168443 Chr.9: 87884582-87884582 on Build GRCh38
rs1028019436 Chr.9: 87884584-87884584 on Build GRCh38
rs557708519 Chr.9: 87884627-87884627 on Build GRCh38
rs752106593 Chr.9: 87884616-87884616 on Build GRCh38
rs1052861554 Chr.9: 87884490-87884490 on Build GRCh38
rs7034636 Chr.9: 87884506-87884506 on Build GRCh38
rs761189997 Chr.9: 87884680-87884680 on Build GRCh38
rs779029637 Chr.9: 87884667-87884667 on Build GRCh38
rs7846811 Chr.9: 87884611-87884611 on Build GRCh38
rs199885345 Chr.9: 87884665-87884665 on Build GRCh38
rs747097121 Chr.9: 87884556-87884556 on Build GRCh38
rs768527477 Chr.9: 87884672-87884672 on Build GRCh38
rs939293679 Chr.9: 87884497-87884497 on Build GRCh38
rs746940066 Chr.9: 87884671-87884671 on Build GRCh38
rs755167389 Chr.9: 87884626-87884626 on Build GRCh38
rs1011414885 Chr.9: 87884509-87884509 on Build GRCh38
rs772719716 Chr.9: 87884696-87884696 on Build GRCh38
rs962261613 Chr.9: 87884624-87884624 on Build GRCh38
rs748176371 Chr.9: 87884668-87884668 on Build GRCh38

View Full Product Details