Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73193055-73193312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG1
Entrez Gene ID
9382
Gene Name
component of oligomeric golgi complex 1
Gene Aliases
CDG2G, LDLB
UniGene
Hs.103555
Gene Chromosome Location
Chr.17: 73193034-73208507 on Build GRCh38
Transcripts
NM_018714.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377644 c.70A>G p.T24A Chr.17: 73193139-73193139 on Build GRCh38
5981282 c.190G>A p.D64N Chr.17: 73193259-73193259 on Build GRCh38
1385668 c.75T>C p.H25H Chr.17: 73193144-73193144 on Build GRCh38
5972384 c.183G>A p.E61E Chr.17: 73193252-73193252 on Build GRCh38
5951628 c.112C>T p.R38W Chr.17: 73193181-73193181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142719529 Chr.17: 73193127-73193127 on Build GRCh38
rs11544801 Chr.17: 73193203-73193203 on Build GRCh38
rs867679947 Chr.17: 73193094-73193094 on Build GRCh38
rs200144944 Chr.17: 73193089-73193089 on Build GRCh38
rs766694294 Chr.17: 73193232-73193232 on Build GRCh38
rs898687512 Chr.17: 73193093-73193093 on Build GRCh38
rs773015047 Chr.17: 73193082-73193082 on Build GRCh38
rs1032371996 Chr.17: 73193154-73193154 on Build GRCh38
rs775416698 Chr.17: 73193177-73193177 on Build GRCh38
rs937851782 Chr.17: 73193250-73193250 on Build GRCh38
rs755637737 Chr.17: 73193258-73193258 on Build GRCh38
rs886053367 Chr.17: 73193269-73193269 on Build GRCh38
rs540003687 Chr.17: 73193274-73193274 on Build GRCh38
rs532947212 Chr.17: 73193080-73193080 on Build GRCh38
rs745589792 Chr.17: 73193285-73193285 on Build GRCh38
rs529636950 Chr.17: 73193148-73193148 on Build GRCh38
rs1020933843 Chr.17: 73193143-73193143 on Build GRCh38
rs757383278 Chr.17: 73193104-73193104 on Build GRCh38
rs769259757 Chr.17: 73193073-73193073 on Build GRCh38
rs759369353 Chr.17: 73193095-73193095 on Build GRCh38
rs781738369 Chr.17: 73193238-73193238 on Build GRCh38
rs375609831 Chr.17: 73193156-73193156 on Build GRCh38
rs372590378 Chr.17: 73193135-73193135 on Build GRCh38
rs551080000 Chr.17: 73193098-73193098 on Build GRCh38
rs201263432 Chr.17: 73193103-73193103 on Build GRCh38
rs775363228 Chr.17: 73193292-73193292 on Build GRCh38
rs751859972 Chr.17: 73193233-73193233 on Build GRCh38
rs371329302 Chr.17: 73193249-73193249 on Build GRCh38
rs759440785 Chr.17: 73193159-73193159 on Build GRCh38
rs762772074 Chr.17: 73193086-73193086 on Build GRCh38
rs774109257 Chr.17: 73193279-73193279 on Build GRCh38
rs139516400 Chr.17: 73193078-73193078 on Build GRCh38
rs374230801 Chr.17: 73193293-73193293 on Build GRCh38
rs750645078 Chr.17: 73193108-73193108 on Build GRCh38
rs758521897 Chr.17: 73193217-73193217 on Build GRCh38
rs143964203 Chr.17: 73193165-73193165 on Build GRCh38
rs767454152 Chr.17: 73193112-73193112 on Build GRCh38
rs567138668 Chr.17: 73193141-73193141 on Build GRCh38
rs376947332 Chr.17: 73193246-73193246 on Build GRCh38
rs769041804 Chr.17: 73193131-73193131 on Build GRCh38
rs1054929490 Chr.17: 73193281-73193281 on Build GRCh38
rs761810855 Chr.17: 73193182-73193182 on Build GRCh38
rs1048879387 Chr.17: 73193237-73193237 on Build GRCh38
rs976164079 Chr.17: 73193222-73193222 on Build GRCh38
rs751465838 Chr.17: 73193091-73193091 on Build GRCh38
rs569505928 Chr.17: 73193259-73193259 on Build GRCh38
rs371047735 Chr.17: 73193211-73193211 on Build GRCh38
rs527295185 Chr.17: 73193116-73193116 on Build GRCh38
rs747496711 Chr.17: 73193072-73193072 on Build GRCh38
rs375382537 Chr.17: 73193234-73193234 on Build GRCh38
rs944279563 Chr.17: 73193195-73193195 on Build GRCh38
rs771901938 Chr.17: 73193288-73193288 on Build GRCh38
rs745468655 Chr.17: 73193292-73193293 on Build GRCh38
rs770665317 Chr.17: 73193153-73193153 on Build GRCh38
rs550969109 Chr.17: 73193210-73193210 on Build GRCh38
rs896415013 Chr.17: 73193289-73193289 on Build GRCh38
rs1021397224 Chr.17: 73193194-73193194 on Build GRCh38

View Full Product Details