Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 983139-983372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf50
Entrez Gene ID
84310
Gene Name
chromosome 7 open reading frame 50
Gene Aliases
YCR016W
UniGene
Hs.653258
Gene Chromosome Location
Chr.7: 978485-1138291 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Gene Symbol
CYP2W1
Entrez Gene ID
54905
Gene Name
cytochrome P450 family 2 subfamily W member 1
Gene Aliases
-
UniGene
Hs.272795
Gene Chromosome Location
Chr.7: 983157-989640 on Build GRCh38
Transcripts
XM_011515580.2 XM_011515581.2 XM_011515582.2 XM_011515583.2 XM_011515584.2 XM_017012720.1 XM_017012721.1 NM_017781.2 XM_011515440.2 XM_011515441.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7327795 c.120C>G p.L40L Chr.7: 983331-983331 on Build GRCh38
6547938 c.80C>T p.P27L Chr.7: 983291-983291 on Build GRCh38
7295905 c.117G>A p.P39P Chr.7: 983328-983328 on Build GRCh38
4748406 c.58G>A p.A20T Chr.7: 983269-983269 on Build GRCh38
5406106 c.110C>T p.P37L Chr.7: 983321-983321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370163399 Chr.7: 983171-983171 on Build GRCh38
rs758410618 Chr.7: 983260-983260 on Build GRCh38
rs773897187 Chr.7: 983187-983187 on Build GRCh38
rs138178506 Chr.7: 983241-983241 on Build GRCh38
rs1045401688 Chr.7: 983217-983217 on Build GRCh38
rs368403278 Chr.7: 983275-983275 on Build GRCh38
rs779903612 Chr.7: 983288-983288 on Build GRCh38
rs754211099 Chr.7: 983226-983226 on Build GRCh38
rs564567759 Chr.7: 983330-983330 on Build GRCh38
rs751702883 Chr.7: 983269-983269 on Build GRCh38
rs924427369 Chr.7: 983185-983185 on Build GRCh38
rs745432053 Chr.7: 983305-983305 on Build GRCh38
rs747790628 Chr.7: 983322-983322 on Build GRCh38
rs541772352 Chr.7: 983162-983162 on Build GRCh38
rs544975130 Chr.7: 983309-983309 on Build GRCh38
rs944198229 Chr.7: 983156-983156 on Build GRCh38
rs1043438349 Chr.7: 983323-983323 on Build GRCh38
rs531323748 Chr.7: 983300-983300 on Build GRCh38
rs1043470503 Chr.7: 983170-983170 on Build GRCh38
rs1013922228 Chr.7: 983205-983205 on Build GRCh38
rs751575496 Chr.7: 983263-983263 on Build GRCh38
rs749076342 Chr.7: 983165-983165 on Build GRCh38
rs376197360 Chr.7: 983200-983200 on Build GRCh38
rs912806496 Chr.7: 983287-983287 on Build GRCh38
rs372411076 Chr.7: 983332-983332 on Build GRCh38
rs951345172 Chr.7: 983274-983274 on Build GRCh38
rs944266423 Chr.7: 983292-983292 on Build GRCh38
rs900002654 Chr.7: 983163-983163 on Build GRCh38
rs777813773 Chr.7: 983268-983268 on Build GRCh38
rs772820393 Chr.7: 983331-983331 on Build GRCh38
rs1031327277 Chr.7: 983197-983197 on Build GRCh38
rs982840128 Chr.7: 983283-983283 on Build GRCh38
rs771722337 Chr.7: 983190-983190 on Build GRCh38
rs562557567 Chr.7: 983299-983299 on Build GRCh38
rs543663619 Chr.7: 983284-983284 on Build GRCh38
rs759843847 Chr.7: 983230-983230 on Build GRCh38
rs777039804 Chr.7: 983335-983335 on Build GRCh38
rs374393317 Chr.7: 983343-983343 on Build GRCh38
rs769704073 Chr.7: 983199-983199 on Build GRCh38
rs929058162 Chr.7: 983271-983271 on Build GRCh38
rs200956100 Chr.7: 983206-983206 on Build GRCh38
rs368456211 Chr.7: 983354-983354 on Build GRCh38
rs1006366537 Chr.7: 983342-983342 on Build GRCh38
rs561461437 Chr.7: 983168-983168 on Build GRCh38
rs999171569 Chr.7: 983178-983178 on Build GRCh38
rs947576204 Chr.7: 983310-983310 on Build GRCh38
rs761366680 Chr.7: 983189-983189 on Build GRCh38

View Full Product Details