Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 73242807-73243080 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf80
Entrez Gene ID
55028
Gene Name
chromosome 17 open reading frame 80
Gene Aliases
HLC-8, MIG3, SPEP1
UniGene
Hs.12929
Gene Chromosome Location
Chr.17: 73232233-73248959 on Build GRCh38
Transcripts
NM_001100621.2 NM_001100622.2 NM_001288770.1 NM_001288771.1 NM_017941.5 NR_110105.1 XM_005257487.3 XM_006721966.3 XM_011524961.1 XM_011524962.2 XM_017024806.1 XM_011525115.2

Gene Symbol
CPSF4L
Entrez Gene ID
642843
Gene Name
cleavage and polyadenylation specific factor 4 like
Gene Aliases
-
UniGene
Hs.534707
Gene Chromosome Location
Chr.17: 73242343-73263778 on Build GRCh38
Transcripts
NM_001100621.2 NM_001100622.2 NM_001288770.1 NM_001288771.1 NM_017941.5 NR_110105.1 XM_005257487.3 XM_006721966.3 XM_011524961.1 XM_011524962.2 XM_017024806.1 XM_011525115.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385688 c.1673C>T p.T558M Chr.17: 73242947-73242947 on Build GRCh38
6259778 c.1622-1G>T p.? Chr.17: 73242895-73242895 on Build GRCh38
6259777 c.1622-1G>T p.? Chr.17: 73242895-73242895 on Build GRCh38
4470174 c.1644C>T p.T548T Chr.17: 73242918-73242918 on Build GRCh38
6405496 c.1667G>A p.G556D Chr.17: 73242941-73242941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749425083 Chr.17: 73242865-73242866 on Build GRCh38
rs145623986 Chr.17: 73242997-73242997 on Build GRCh38
rs755583038 Chr.17: 73242844-73242844 on Build GRCh38
rs377399920 Chr.17: 73243016-73243016 on Build GRCh38
rs760752781 Chr.17: 73242876-73242876 on Build GRCh38
rs753087413 Chr.17: 73243010-73243010 on Build GRCh38
rs778789967 Chr.17: 73242921-73242924 on Build GRCh38
rs998143243 Chr.17: 73242900-73242900 on Build GRCh38
rs991571850 Chr.17: 73243001-73243001 on Build GRCh38
rs377304310 Chr.17: 73243047-73243047 on Build GRCh38
rs771958350 Chr.17: 73243001-73243004 on Build GRCh38
rs745626577 Chr.17: 73242862-73242862 on Build GRCh38
rs533016680 Chr.17: 73242859-73242859 on Build GRCh38
rs778397466 Chr.17: 73242947-73242947 on Build GRCh38
rs753193656 Chr.17: 73242929-73242929 on Build GRCh38
rs948754554 Chr.17: 73242839-73242839 on Build GRCh38
rs748816323 Chr.17: 73242938-73242938 on Build GRCh38
rs201221175 Chr.17: 73242936-73242936 on Build GRCh38
rs139908935 Chr.17: 73242954-73242954 on Build GRCh38
rs369923395 Chr.17: 73242959-73242959 on Build GRCh38
rs547305905 Chr.17: 73242911-73242911 on Build GRCh38
rs2270728 Chr.17: 73243051-73243051 on Build GRCh38
rs748436815 Chr.17: 73242984-73242984 on Build GRCh38
rs16977551 Chr.17: 73242837-73242837 on Build GRCh38
rs113017423 Chr.17: 73242998-73242998 on Build GRCh38
rs1011740691 Chr.17: 73242925-73242925 on Build GRCh38
rs756838424 Chr.17: 73242945-73242945 on Build GRCh38
rs775563182 Chr.17: 73242871-73242871 on Build GRCh38
rs963960626 Chr.17: 73242886-73242886 on Build GRCh38
rs757374605 Chr.17: 73242909-73242911 on Build GRCh38
rs2270726 Chr.17: 73242861-73242861 on Build GRCh38
rs200738758 Chr.17: 73242937-73242937 on Build GRCh38
rs951224562 Chr.17: 73242926-73242926 on Build GRCh38
rs1053095830 Chr.17: 73242878-73242878 on Build GRCh38
rs745953557 Chr.17: 73242950-73242950 on Build GRCh38
rs375628442 Chr.17: 73242927-73242927 on Build GRCh38
rs1566290 Chr.17: 73242948-73242948 on Build GRCh38
rs748869314 Chr.17: 73242851-73242851 on Build GRCh38
rs149730486 Chr.17: 73242973-73242973 on Build GRCh38
rs775379784 Chr.17: 73243048-73243049 on Build GRCh38
rs373380685 Chr.17: 73242971-73242971 on Build GRCh38
rs766534337 Chr.17: 73242914-73242914 on Build GRCh38
rs778543122 Chr.17: 73243048-73243048 on Build GRCh38
rs374965719 Chr.17: 73242979-73242979 on Build GRCh38
rs778175926 Chr.17: 73242855-73242856 on Build GRCh38
rs1015344016 Chr.17: 73243045-73243045 on Build GRCh38
rs773430804 Chr.17: 73242890-73242890 on Build GRCh38
rs767720806 Chr.17: 73243004-73243004 on Build GRCh38
rs764560121 Chr.17: 73243027-73243027 on Build GRCh38
rs560065468 Chr.17: 73242868-73242868 on Build GRCh38
rs2270727 Chr.17: 73242877-73242877 on Build GRCh38
rs751860675 Chr.17: 73242919-73242919 on Build GRCh38
rs9896708 Chr.17: 73243013-73243013 on Build GRCh38

View Full Product Details