Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110327907-110328140 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2A2
Entrez Gene ID
488
Gene Name
ATPase sarcoplasmic/endoplasmic reticulum Ca2+ transporting 2
Gene Aliases
ATP2B, DAR, DD, SERCA2
UniGene
Hs.506759
Gene Chromosome Location
Chr.12: 110281227-110351093 on Build GRCh38
Transcripts
NM_001681.3 NM_170665.3 XM_005253888.2 XM_011538402.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6995734 c.1049C>T p.S350L Chr.12: 110327971-110327971 on Build GRCh38
7202074 c.1001G>T p.R334L Chr.12: 110327923-110327923 on Build GRCh38
6607511 c.1011G>A p.P337P Chr.12: 110327933-110327933 on Build GRCh38
5527112 c.1013C>T p.S338F Chr.12: 110327935-110327935 on Build GRCh38
5051321 c.1063A>G p.T355A Chr.12: 110327985-110327985 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761449131 Chr.12: 110328029-110328029 on Build GRCh38
rs776147205 Chr.12: 110328002-110328002 on Build GRCh38
rs747319498 Chr.12: 110327957-110327957 on Build GRCh38
rs965119749 Chr.12: 110328065-110328065 on Build GRCh38
rs373177511 Chr.12: 110328043-110328043 on Build GRCh38
rs746331231 Chr.12: 110327933-110327933 on Build GRCh38
rs762827295 Chr.12: 110328044-110328044 on Build GRCh38
rs769893630 Chr.12: 110327966-110327966 on Build GRCh38
rs534083202 Chr.12: 110328104-110328104 on Build GRCh38
rs919268720 Chr.12: 110328045-110328045 on Build GRCh38
rs1049027708 Chr.12: 110328114-110328114 on Build GRCh38
rs866239874 Chr.12: 110327932-110327932 on Build GRCh38
rs764627069 Chr.12: 110328031-110328031 on Build GRCh38
rs968205171 Chr.12: 110327978-110327978 on Build GRCh38
rs754751649 Chr.12: 110328062-110328062 on Build GRCh38
rs1803737 Chr.12: 110327938-110327938 on Build GRCh38
rs768304511 Chr.12: 110327987-110327987 on Build GRCh38
rs779524206 Chr.12: 110327930-110327930 on Build GRCh38
rs751411697 Chr.12: 110328055-110328055 on Build GRCh38
rs766366890 Chr.12: 110328049-110328049 on Build GRCh38
rs986287811 Chr.12: 110328012-110328012 on Build GRCh38
rs772529432 Chr.12: 110327936-110327936 on Build GRCh38
rs775857746 Chr.12: 110327951-110327951 on Build GRCh38
rs192091032 Chr.12: 110328072-110328072 on Build GRCh38

View Full Product Details