Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 45307598-45307823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKM
Entrez Gene ID
1158
Gene Name
creatine kinase, M-type
Gene Aliases
CKMM, M-CK
UniGene
Hs.334347
Gene Chromosome Location
Chr.19: 45306413-45322977 on Build GRCh38
Transcripts
NM_001824.4 XM_017026240.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7361383 c.805C>A p.H269N Chr.19: 45307623-45307623 on Build GRCh38
4672540 c.778-1G>A p.? Chr.19: 45307651-45307651 on Build GRCh38
3103982 c.817T>C p.W273R Chr.19: 45307611-45307611 on Build GRCh38
998172 c.789C>A p.I263I Chr.19: 45307639-45307639 on Build GRCh38
998171 c.803G>A p.G268D Chr.19: 45307625-45307625 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747623819 Chr.19: 45307627-45307627 on Build GRCh38
rs985340658 Chr.19: 45307771-45307771 on Build GRCh38
rs767676171 Chr.19: 45307655-45307655 on Build GRCh38
rs537324872 Chr.19: 45307705-45307705 on Build GRCh38
rs149187786 Chr.19: 45307708-45307708 on Build GRCh38
rs191206227 Chr.19: 45307759-45307759 on Build GRCh38
rs769629174 Chr.19: 45307702-45307702 on Build GRCh38
rs780521813 Chr.19: 45307693-45307693 on Build GRCh38
rs565328498 Chr.19: 45307784-45307784 on Build GRCh38
rs771408722 Chr.19: 45307645-45307645 on Build GRCh38
rs772900129 Chr.19: 45307629-45307629 on Build GRCh38
rs1027362248 Chr.19: 45307682-45307682 on Build GRCh38
rs769481477 Chr.19: 45307691-45307692 on Build GRCh38
rs774709360 Chr.19: 45307649-45307649 on Build GRCh38
rs751322808 Chr.19: 45307684-45307684 on Build GRCh38
rs763504810 Chr.19: 45307642-45307642 on Build GRCh38
rs752946274 Chr.19: 45307659-45307659 on Build GRCh38
rs8104850 Chr.19: 45307685-45307685 on Build GRCh38
rs763366065 Chr.19: 45307670-45307670 on Build GRCh38
rs759831359 Chr.19: 45307651-45307651 on Build GRCh38
rs576465646 Chr.19: 45307750-45307750 on Build GRCh38
rs966339713 Chr.19: 45307774-45307774 on Build GRCh38
rs760826961 Chr.19: 45307619-45307619 on Build GRCh38
rs760487207 Chr.19: 45307634-45307634 on Build GRCh38
rs779872868 Chr.19: 45307683-45307683 on Build GRCh38
rs763019668 Chr.19: 45307693-45307694 on Build GRCh38
rs747780777 Chr.19: 45307697-45307697 on Build GRCh38
rs111597734 Chr.19: 45307653-45307653 on Build GRCh38
rs758348088 Chr.19: 45307681-45307681 on Build GRCh38
rs912481095 Chr.19: 45307798-45307798 on Build GRCh38
rs753918375 Chr.19: 45307674-45307674 on Build GRCh38
rs138693691 Chr.19: 45307628-45307628 on Build GRCh38
rs775013601 Chr.19: 45307622-45307622 on Build GRCh38
rs781176919 Chr.19: 45307696-45307696 on Build GRCh38
rs757580716 Chr.19: 45307646-45307646 on Build GRCh38

View Full Product Details