Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 53046733-53046962 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDHD1
Entrez Gene ID
80821
Gene Name
DDHD domain containing 1
Gene Aliases
PA-PLA1, PAPLA1, SPG28
UniGene
Hs.125525
Gene Chromosome Location
Chr.14: 53036740-53153328 on Build GRCh38
Transcripts
NM_001160147.1 NM_001160148.1 NM_030637.2 XM_005268102.2 XM_005268103.2 XM_005268105.2 XM_011537188.2 XM_011537189.2 XM_017021668.1 XM_017021669.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6007976 c.2564T>A p.F855Y Chr.14: 53046823-53046823 on Build GRCh38
6641289 c.2576G>A p.R859H Chr.14: 53046895-53046895 on Build GRCh38
1147327 c.2622G>T p.L874F Chr.14: 53046849-53046849 on Build GRCh38
4453137 c.2495A>C p.Y832S Chr.14: 53046892-53046892 on Build GRCh38
6641283 c.2596T>C p.S866P Chr.14: 53046875-53046875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376026633 Chr.14: 53046771-53046771 on Build GRCh38
rs752058566 Chr.14: 53046841-53046841 on Build GRCh38
rs763731870 Chr.14: 53046808-53046808 on Build GRCh38
rs3037072 Chr.14: 53046764-53046765 on Build GRCh38
rs774311230 Chr.14: 53046789-53046789 on Build GRCh38
rs989346515 Chr.14: 53046797-53046797 on Build GRCh38
rs911729593 Chr.14: 53046821-53046821 on Build GRCh38
rs749564580 Chr.14: 53046877-53046877 on Build GRCh38
rs199980471 Chr.14: 53046765-53046765 on Build GRCh38
rs757075549 Chr.14: 53046882-53046882 on Build GRCh38
rs10599915 Chr.14: 53046761-53046762 on Build GRCh38
rs760306971 Chr.14: 53046806-53046806 on Build GRCh38
rs15873 Chr.14: 53046832-53046832 on Build GRCh38
rs752718428 Chr.14: 53046869-53046869 on Build GRCh38
rs371021311 Chr.14: 53046788-53046788 on Build GRCh38
rs774873200 Chr.14: 53046804-53046804 on Build GRCh38
rs141960546 Chr.14: 53046817-53046817 on Build GRCh38
rs749084227 Chr.14: 53046787-53046787 on Build GRCh38
rs755453442 Chr.14: 53046845-53046845 on Build GRCh38
rs539889476 Chr.14: 53046930-53046930 on Build GRCh38
rs202164262 Chr.14: 53046803-53046803 on Build GRCh38
rs748343890 Chr.14: 53046778-53046778 on Build GRCh38
rs1036525672 Chr.14: 53046896-53046896 on Build GRCh38
rs184128645 Chr.14: 53046931-53046931 on Build GRCh38
rs112753891 Chr.14: 53046887-53046887 on Build GRCh38
rs33960665 Chr.14: 53046760-53046761 on Build GRCh38
rs192934516 Chr.14: 53046782-53046782 on Build GRCh38
rs112305277 Chr.14: 53046762-53046763 on Build GRCh38
rs397712234 Chr.14: 53046765-53046766 on Build GRCh38
rs767567231 Chr.14: 53046863-53046863 on Build GRCh38
rs1032973840 Chr.14: 53046766-53046766 on Build GRCh38
rs527804109 Chr.14: 53046785-53046785 on Build GRCh38
rs570950752 Chr.14: 53046830-53046830 on Build GRCh38
rs766762679 Chr.14: 53046839-53046839 on Build GRCh38
rs957406664 Chr.14: 53046768-53046768 on Build GRCh38
rs944838879 Chr.14: 53046915-53046915 on Build GRCh38
rs374202355 Chr.14: 53046822-53046822 on Build GRCh38
rs973996851 Chr.14: 53046762-53046762 on Build GRCh38
rs756324099 Chr.14: 53046873-53046873 on Build GRCh38
rs778120921 Chr.14: 53046876-53046876 on Build GRCh38
rs866490429 Chr.14: 53046837-53046837 on Build GRCh38
rs781154518 Chr.14: 53046773-53046773 on Build GRCh38

View Full Product Details