Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53015375-53015600 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERVV-1
Entrez Gene ID
147664
Gene Name
endogenous retrovirus group V member 1
Gene Aliases
ENVV1, HERV-V1
UniGene
Hs.44329
Gene Chromosome Location
Chr.19: 53014091-53016580 on Build GRCh38
Transcripts
NM_152473.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861888 c.1309G>T p.D437Y Chr.19: 53015399-53015399 on Build GRCh38
4953884 c.1319G>T p.G440V Chr.19: 53015409-53015409 on Build GRCh38
5792539 c.1345G>T p.A449S Chr.19: 53015435-53015435 on Build GRCh38
6987934 c.1320G>T p.G440G Chr.19: 53015410-53015410 on Build GRCh38
6341990 c.1326T>C p.I442I Chr.19: 53015416-53015416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs899378165 Chr.19: 53015445-53015445 on Build GRCh38
rs556877432 Chr.19: 53015410-53015410 on Build GRCh38
rs537563356 Chr.19: 53015457-53015457 on Build GRCh38
rs967939912 Chr.19: 53015561-53015561 on Build GRCh38
rs570563995 Chr.19: 53015447-53015447 on Build GRCh38
rs1015710925 Chr.19: 53015401-53015401 on Build GRCh38
rs149606721 Chr.19: 53015511-53015511 on Build GRCh38
rs1650925 Chr.19: 53015494-53015494 on Build GRCh38
rs760872150 Chr.19: 53015554-53015554 on Build GRCh38
rs775115851 Chr.19: 53015575-53015575 on Build GRCh38
rs990927351 Chr.19: 53015548-53015548 on Build GRCh38
rs977960065 Chr.19: 53015565-53015565 on Build GRCh38
rs575051596 Chr.19: 53015464-53015464 on Build GRCh38
rs959348393 Chr.19: 53015541-53015541 on Build GRCh38
rs775758077 Chr.19: 53015530-53015530 on Build GRCh38

View Full Product Details