Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123693608-123693879 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANXA13
Entrez Gene ID
312
Gene Name
annexin A13
Gene Aliases
ANX13, ISA
UniGene
Hs.181107
Gene Chromosome Location
Chr.8: 123680794-123737426 on Build GRCh38
Transcripts
NM_001003954.2 NM_004306.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7142296 c.601C>A p.R201S Chr.8: 123693773-123693773 on Build GRCh38
6837312 c.640C>A p.Q214K Chr.8: 123693734-123693734 on Build GRCh38
1095806 c.651A>C p.K217N Chr.8: 123693723-123693723 on Build GRCh38
3432095 c.628G>T p.D210Y Chr.8: 123693746-123693746 on Build GRCh38
3896580 c.626A>G p.K209R Chr.8: 123693748-123693748 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs773180630 Chr.8: 123693695-123693695 on Build GRCh38
rs530627306 Chr.8: 123693696-123693696 on Build GRCh38
rs528363634 Chr.8: 123693652-123693652 on Build GRCh38
rs747905174 Chr.8: 123693683-123693683 on Build GRCh38
rs532899750 Chr.8: 123693773-123693773 on Build GRCh38
rs149725897 Chr.8: 123693779-123693779 on Build GRCh38
rs913476496 Chr.8: 123693732-123693732 on Build GRCh38
rs868002260 Chr.8: 123693767-123693767 on Build GRCh38
rs753266845 Chr.8: 123693738-123693738 on Build GRCh38
rs776512903 Chr.8: 123693712-123693712 on Build GRCh38
rs1014824122 Chr.8: 123693843-123693843 on Build GRCh38
rs75881016 Chr.8: 123693850-123693850 on Build GRCh38
rs763493417 Chr.8: 123693740-123693740 on Build GRCh38
rs561910888 Chr.8: 123693670-123693670 on Build GRCh38
rs145382855 Chr.8: 123693719-123693719 on Build GRCh38
rs185357145 Chr.8: 123693794-123693794 on Build GRCh38
rs756221312 Chr.8: 123693755-123693755 on Build GRCh38
rs200689921 Chr.8: 123693669-123693669 on Build GRCh38
rs376084774 Chr.8: 123693772-123693772 on Build GRCh38
rs770991731 Chr.8: 123693820-123693820 on Build GRCh38
rs972270318 Chr.8: 123693756-123693756 on Build GRCh38
rs35759790 Chr.8: 123693777-123693777 on Build GRCh38
rs778338256 Chr.8: 123693792-123693792 on Build GRCh38
rs771967926 Chr.8: 123693691-123693691 on Build GRCh38
rs776942718 Chr.8: 123693829-123693829 on Build GRCh38
rs748065321 Chr.8: 123693793-123693793 on Build GRCh38
rs12056704 Chr.8: 123693807-123693807 on Build GRCh38
rs778941952 Chr.8: 123693678-123693678 on Build GRCh38
rs765618274 Chr.8: 123693722-123693722 on Build GRCh38
rs764441564 Chr.8: 123693768-123693768 on Build GRCh38
rs201263416 Chr.8: 123693842-123693843 on Build GRCh38
rs916083545 Chr.8: 123693816-123693816 on Build GRCh38
rs779420642 Chr.8: 123693660-123693660 on Build GRCh38
rs550213175 Chr.8: 123693711-123693711 on Build GRCh38
rs887577955 Chr.8: 123693833-123693833 on Build GRCh38
rs746810025 Chr.8: 123693818-123693818 on Build GRCh38
rs777696663 Chr.8: 123693804-123693804 on Build GRCh38

View Full Product Details