Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 73726525-73726758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMSAN1
Entrez Gene ID
91748
Gene Name
ELM2 and Myb/SANT domain containing 1
Gene Aliases
C14orf117, C14orf43, LSR68, MIDEAS, c14_5541
UniGene
Hs.594157
Gene Chromosome Location
Chr.14: 73715122-73790285 on Build GRCh38
Transcripts
NM_001043318.2 NM_194278.3 XM_005268204.1 XM_005268205.1 XM_005268206.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1748975 c.2319G>A p.L773L Chr.14: 73726694-73726694 on Build GRCh38
6869755 c.2330G>T p.C777F Chr.14: 73726683-73726683 on Build GRCh38
2137328 c.2354G>A p.G785D Chr.14: 73726659-73726659 on Build GRCh38
5049140 c.2336G>T p.S779I Chr.14: 73726677-73726677 on Build GRCh38
957677 c.2389G>A p.E797K Chr.14: 73726624-73726624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781668774 Chr.14: 73726699-73726699 on Build GRCh38
rs770225341 Chr.14: 73726720-73726720 on Build GRCh38
rs758461082 Chr.14: 73726681-73726681 on Build GRCh38
rs776026107 Chr.14: 73726722-73726722 on Build GRCh38
rs752738730 Chr.14: 73726666-73726666 on Build GRCh38
rs375272031 Chr.14: 73726625-73726625 on Build GRCh38
rs992605088 Chr.14: 73726691-73726691 on Build GRCh38
rs887508 Chr.14: 73726724-73726724 on Build GRCh38
rs771554444 Chr.14: 73726723-73726723 on Build GRCh38
rs113920409 Chr.14: 73726579-73726579 on Build GRCh38
rs768256400 Chr.14: 73726555-73726555 on Build GRCh38
rs760474043 Chr.14: 73726609-73726609 on Build GRCh38
rs868709021 Chr.14: 73726659-73726659 on Build GRCh38
rs767109475 Chr.14: 73726585-73726585 on Build GRCh38
rs201907899 Chr.14: 73726572-73726572 on Build GRCh38
rs577063865 Chr.14: 73726687-73726687 on Build GRCh38
rs765402633 Chr.14: 73726657-73726657 on Build GRCh38
rs902800984 Chr.14: 73726591-73726591 on Build GRCh38
rs367842372 Chr.14: 73726711-73726711 on Build GRCh38
rs999745170 Chr.14: 73726610-73726610 on Build GRCh38
rs754839238 Chr.14: 73726651-73726651 on Build GRCh38
rs1035163604 Chr.14: 73726664-73726664 on Build GRCh38
rs746430105 Chr.14: 73726717-73726717 on Build GRCh38
rs1003051881 Chr.14: 73726663-73726663 on Build GRCh38
rs148101731 Chr.14: 73726727-73726727 on Build GRCh38
rs772786760 Chr.14: 73726592-73726592 on Build GRCh38
rs201285016 Chr.14: 73726688-73726688 on Build GRCh38
rs753839319 Chr.14: 73726634-73726634 on Build GRCh38
rs773713188 Chr.14: 73726568-73726568 on Build GRCh38
rs971130453 Chr.14: 73726589-73726589 on Build GRCh38
rs377105370 Chr.14: 73726642-73726642 on Build GRCh38

View Full Product Details