Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 17244955-17245184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT7
Entrez Gene ID
29883
Gene Name
CCR4-NOT transcription complex subunit 7
Gene Aliases
CAF-1, CAF1, Caf1a, hCAF-1
UniGene
Hs.587465
Gene Chromosome Location
Chr.8: 17226030-17246909 on Build GRCh38
Transcripts
NM_001322087.1 NM_001322088.1 NM_001322089.1 NM_001322090.1 NM_001322091.1 NM_001322092.1 NM_001322093.1 NM_001322094.1 NM_001322095.1 NM_001322096.1 NM_001322097.1 NM_001322098.1 NM_001322099.1 NM_001322100.1 NM_013354.6 NM_054026.3 XM_005273481.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6631556 c.108C>T p.Y36Y Chr.8: 17245045-17245045 on Build GRCh38
6264492 c.87A>G p.Q29Q Chr.8: 17245066-17245066 on Build GRCh38
6455673 c.65A>T p.E22V Chr.8: 17245088-17245088 on Build GRCh38
1496863 c.27C>A p.S9R Chr.8: 17245126-17245126 on Build GRCh38
3951478 c.1A>G p.M1V Chr.8: 17245152-17245152 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200983574 Chr.8: 17245081-17245081 on Build GRCh38
rs762916263 Chr.8: 17245126-17245126 on Build GRCh38
rs187178007 Chr.8: 17245009-17245009 on Build GRCh38
rs369445114 Chr.8: 17245083-17245083 on Build GRCh38
rs760446086 Chr.8: 17244995-17244995 on Build GRCh38
rs746540566 Chr.8: 17245084-17245084 on Build GRCh38
rs766330962 Chr.8: 17245139-17245139 on Build GRCh38
rs376194907 Chr.8: 17245144-17245144 on Build GRCh38
rs930242144 Chr.8: 17245104-17245104 on Build GRCh38
rs183346094 Chr.8: 17245001-17245001 on Build GRCh38
rs751586478 Chr.8: 17245140-17245140 on Build GRCh38
rs375278542 Chr.8: 17245058-17245058 on Build GRCh38
rs778137902 Chr.8: 17245070-17245070 on Build GRCh38
rs753027730 Chr.8: 17245147-17245147 on Build GRCh38
rs767567772 Chr.8: 17245145-17245145 on Build GRCh38
rs148569654 Chr.8: 17245044-17245044 on Build GRCh38
rs757790929 Chr.8: 17245161-17245161 on Build GRCh38
rs749702643 Chr.8: 17245156-17245156 on Build GRCh38
rs768140191 Chr.8: 17245091-17245091 on Build GRCh38
rs970479963 Chr.8: 17245116-17245116 on Build GRCh38
rs765021645 Chr.8: 17245008-17245008 on Build GRCh38
rs776354768 Chr.8: 17245092-17245092 on Build GRCh38
rs755150086 Chr.8: 17245026-17245026 on Build GRCh38
rs373257623 Chr.8: 17245099-17245099 on Build GRCh38
rs376624089 Chr.8: 17245154-17245154 on Build GRCh38
rs753527852 Chr.8: 17245004-17245004 on Build GRCh38
rs369921308 Chr.8: 17245153-17245153 on Build GRCh38
rs549878313 Chr.8: 17245111-17245111 on Build GRCh38
rs781083959 Chr.8: 17245052-17245052 on Build GRCh38
rs775261886 Chr.8: 17244989-17244989 on Build GRCh38
rs766492823 Chr.8: 17245021-17245021 on Build GRCh38
rs142912321 Chr.8: 17245097-17245097 on Build GRCh38
rs959574445 Chr.8: 17245030-17245030 on Build GRCh38
rs192001497 Chr.8: 17245022-17245022 on Build GRCh38
rs527875297 Chr.8: 17245006-17245006 on Build GRCh38

View Full Product Details