Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 99896952-99897225 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EML1
Entrez Gene ID
2009
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
ELP79, EMAP, EMAPL, HuEMAP
UniGene
Hs.12451
Gene Chromosome Location
Chr.14: 99737699-99942060 on Build GRCh38
Transcripts
NM_001008707.1 NM_004434.2 XM_005267397.1 XM_005267398.3 XM_005267399.4 XM_005267400.4 XM_011536540.2 XM_011536541.1 XM_011536542.2 XM_017021074.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
949263 c.810A>G p.A270A Chr.14: 99897220-99897220 on Build GRCh38
3401113 c.761G>A p.R254H Chr.14: 99897171-99897171 on Build GRCh38
6324218 c.785C>T p.T262M Chr.14: 99897195-99897195 on Build GRCh38
5679464 c.771G>T p.L257L Chr.14: 99897181-99897181 on Build GRCh38
4049040 c.782C>T p.P261L Chr.14: 99897192-99897192 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989486436 Chr.14: 99897038-99897038 on Build GRCh38
rs199917254 Chr.14: 99897096-99897096 on Build GRCh38
rs754695102 Chr.14: 99897005-99897005 on Build GRCh38
rs922601466 Chr.14: 99897044-99897044 on Build GRCh38
rs956298405 Chr.14: 99897015-99897015 on Build GRCh38
rs939200582 Chr.14: 99897196-99897196 on Build GRCh38
rs758244572 Chr.14: 99897172-99897172 on Build GRCh38
rs777662441 Chr.14: 99897178-99897178 on Build GRCh38
rs368509823 Chr.14: 99897193-99897193 on Build GRCh38
rs777466555 Chr.14: 99897147-99897147 on Build GRCh38
rs17099193 Chr.14: 99897076-99897076 on Build GRCh38
rs1037525338 Chr.14: 99897195-99897195 on Build GRCh38
rs778702524 Chr.14: 99897155-99897155 on Build GRCh38
rs766018312 Chr.14: 99897117-99897117 on Build GRCh38
rs747890251 Chr.14: 99897157-99897157 on Build GRCh38
rs760160905 Chr.14: 99897123-99897123 on Build GRCh38
rs755861337 Chr.14: 99897144-99897144 on Build GRCh38
rs753481337 Chr.14: 99897151-99897151 on Build GRCh38
rs1033170126 Chr.14: 99897012-99897012 on Build GRCh38
rs767103627 Chr.14: 99897006-99897006 on Build GRCh38
rs752157174 Chr.14: 99897115-99897115 on Build GRCh38
rs989177866 Chr.14: 99897072-99897072 on Build GRCh38
rs970159654 Chr.14: 99897000-99897000 on Build GRCh38
rs146448225 Chr.14: 99897205-99897205 on Build GRCh38
rs753790779 Chr.14: 99897199-99897199 on Build GRCh38
rs1017313299 Chr.14: 99896979-99896979 on Build GRCh38
rs941458330 Chr.14: 99897121-99897121 on Build GRCh38
rs201705960 Chr.14: 99897105-99897105 on Build GRCh38
rs17652784 Chr.14: 99896999-99896999 on Build GRCh38
rs751798883 Chr.14: 99897066-99897066 on Build GRCh38
rs755656032 Chr.14: 99897114-99897114 on Build GRCh38
rs766937956 Chr.14: 99897114-99897115 on Build GRCh38
rs746986316 Chr.14: 99897192-99897192 on Build GRCh38
rs886037936 Chr.14: 99897194-99897194 on Build GRCh38

View Full Product Details