Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 10290561-10290792 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CMBL
Entrez Gene ID
134147
Gene Name
carboxymethylenebutenolidase homolog
Gene Aliases
JS-1
UniGene
Hs.192586
Gene Chromosome Location
Chr.5: 10277595-10308056 on Build GRCh38
Transcripts
NM_138809.3 XM_017009034.1 XM_017009035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
735157 c.83A>G p.E28G Chr.5: 10290680-10290680 on Build GRCh38
2990092 c.59G>T p.G20V Chr.5: 10290704-10290704 on Build GRCh38
5987188 c.76C>G p.Q26E Chr.5: 10290687-10290687 on Build GRCh38
735156 c.34A>G p.I12V Chr.5: 10290729-10290729 on Build GRCh38
7141330 c.141C>A p.V47V Chr.5: 10290622-10290622 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955945391 Chr.5: 10290681-10290681 on Build GRCh38
rs113456062 Chr.5: 10290720-10290720 on Build GRCh38
rs975622373 Chr.5: 10290651-10290651 on Build GRCh38
rs929375859 Chr.5: 10290753-10290753 on Build GRCh38
rs755532552 Chr.5: 10290764-10290764 on Build GRCh38
rs781421726 Chr.5: 10290721-10290721 on Build GRCh38
rs765600000 Chr.5: 10290695-10290695 on Build GRCh38
rs146175625 Chr.5: 10290649-10290649 on Build GRCh38
rs372804433 Chr.5: 10290728-10290728 on Build GRCh38
rs777739414 Chr.5: 10290727-10290727 on Build GRCh38
rs369783862 Chr.5: 10290648-10290648 on Build GRCh38
rs770071059 Chr.5: 10290725-10290725 on Build GRCh38
rs967514119 Chr.5: 10290600-10290600 on Build GRCh38
rs770734908 Chr.5: 10290729-10290729 on Build GRCh38
rs562374337 Chr.5: 10290640-10290640 on Build GRCh38
rs148288379 Chr.5: 10290682-10290682 on Build GRCh38
rs758977998 Chr.5: 10290698-10290698 on Build GRCh38
rs752226148 Chr.5: 10290630-10290631 on Build GRCh38
rs757744396 Chr.5: 10290698-10290703 on Build GRCh38
rs773078898 Chr.5: 10290625-10290625 on Build GRCh38
rs771628700 Chr.5: 10290736-10290736 on Build GRCh38
rs923305448 Chr.5: 10290738-10290738 on Build GRCh38
rs374486111 Chr.5: 10290675-10290675 on Build GRCh38
rs914052806 Chr.5: 10290690-10290690 on Build GRCh38
rs760306446 Chr.5: 10290688-10290688 on Build GRCh38
rs763697274 Chr.5: 10290740-10290740 on Build GRCh38
rs769783517 Chr.5: 10290624-10290624 on Build GRCh38
rs766738980 Chr.5: 10290761-10290761 on Build GRCh38
rs531294435 Chr.5: 10290758-10290758 on Build GRCh38
rs140447154 Chr.5: 10290735-10290735 on Build GRCh38
rs748390410 Chr.5: 10290722-10290722 on Build GRCh38
rs377251688 Chr.5: 10290734-10290734 on Build GRCh38
rs752087307 Chr.5: 10290712-10290712 on Build GRCh38
rs755153125 Chr.5: 10290714-10290714 on Build GRCh38
rs143191732 Chr.5: 10290659-10290659 on Build GRCh38
rs750687273 Chr.5: 10290696-10290696 on Build GRCh38
rs781617780 Chr.5: 10290612-10290612 on Build GRCh38
rs773650498 Chr.5: 10290754-10290754 on Build GRCh38
rs763279164 Chr.5: 10290756-10290756 on Build GRCh38
rs553743018 Chr.5: 10290762-10290762 on Build GRCh38
rs201605601 Chr.5: 10290623-10290623 on Build GRCh38

View Full Product Details