Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 93355264-93355491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEUP1
Entrez Gene ID
159989
Gene Name
deuterosome assembly protein 1
Gene Aliases
CCDC67
UniGene
Hs.436625
Gene Chromosome Location
Chr.11: 93329079-93438470 on Build GRCh38
Transcripts
NM_181645.3 XM_005273802.1 XM_011542629.2 XM_011542630.1 XM_011542631.2 XM_011542632.2 XM_011542633.2 XM_011542635.2 XM_011542637.2 XM_017017245.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1200032 c.45G>T p.E15D Chr.11: 93355386-93355386 on Build GRCh38
6271434 c.85G>T p.V29L Chr.11: 93355426-93355426 on Build GRCh38
4669998 c.122G>A p.R41Q Chr.11: 93355463-93355463 on Build GRCh38
2224401 c.121C>T p.R41W Chr.11: 93355462-93355462 on Build GRCh38
2224402 c.136C>T p.R46* Chr.11: 93355477-93355477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765730078 Chr.11: 93355356-93355356 on Build GRCh38
rs773985601 Chr.11: 93355321-93355321 on Build GRCh38
rs774202838 Chr.11: 93355429-93355429 on Build GRCh38
rs532880533 Chr.11: 93355337-93355337 on Build GRCh38
rs775439544 Chr.11: 93355393-93355393 on Build GRCh38
rs376502642 Chr.11: 93355415-93355415 on Build GRCh38
rs373323890 Chr.11: 93355326-93355326 on Build GRCh38
rs747221324 Chr.11: 93355370-93355370 on Build GRCh38
rs764499863 Chr.11: 93355464-93355464 on Build GRCh38
rs201641405 Chr.11: 93355358-93355358 on Build GRCh38
rs773396213 Chr.11: 93355456-93355457 on Build GRCh38
rs761532182 Chr.11: 93355446-93355446 on Build GRCh38
rs924457929 Chr.11: 93355378-93355378 on Build GRCh38
rs760209195 Chr.11: 93355457-93355457 on Build GRCh38
rs768491574 Chr.11: 93355422-93355422 on Build GRCh38
rs758750223 Chr.11: 93355463-93355463 on Build GRCh38
rs769641235 Chr.11: 93355389-93355389 on Build GRCh38
rs200488619 Chr.11: 93355462-93355462 on Build GRCh38
rs781219200 Chr.11: 93355380-93355380 on Build GRCh38
rs757377379 Chr.11: 93355376-93355376 on Build GRCh38
rs201790053 Chr.11: 93355449-93355449 on Build GRCh38
rs78902781 Chr.11: 93355341-93355341 on Build GRCh38
rs957289196 Chr.11: 93355401-93355401 on Build GRCh38
rs201030952 Chr.11: 93355366-93355366 on Build GRCh38
rs968942111 Chr.11: 93355360-93355360 on Build GRCh38
rs758725297 Chr.11: 93355362-93355362 on Build GRCh38
rs745908412 Chr.11: 93355384-93355384 on Build GRCh38
rs530193317 Chr.11: 93355347-93355347 on Build GRCh38
rs772875843 Chr.11: 93355450-93355450 on Build GRCh38
rs769955257 Chr.11: 93355376-93355378 on Build GRCh38

View Full Product Details