Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30552398-30552669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNL1
Entrez Gene ID
2794
Gene Name
G protein nucleolar 1 (putative)
Gene Aliases
HSR1
UniGene
Hs.118354
Gene Chromosome Location
Chr.6: 30541377-30557594 on Build GRCh38
Transcripts
NM_005275.3 XM_005249015.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391629 c.1033G>A p.A345T Chr.6: 30552533-30552533 on Build GRCh38
5737526 c.923G>A p.R308Q Chr.6: 30552643-30552643 on Build GRCh38
6409071 c.976G>A p.E326K Chr.6: 30552590-30552590 on Build GRCh38
5885457 c.930G>C p.K310N Chr.6: 30552636-30552636 on Build GRCh38
3624256 c.993G>A p.E331E Chr.6: 30552573-30552573 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749555843 Chr.6: 30552555-30552555 on Build GRCh38
rs142734504 Chr.6: 30552628-30552628 on Build GRCh38
rs112012560 Chr.6: 30552456-30552456 on Build GRCh38
rs545644145 Chr.6: 30552476-30552476 on Build GRCh38
rs760959959 Chr.6: 30552591-30552591 on Build GRCh38
rs142398523 Chr.6: 30552590-30552590 on Build GRCh38
rs191921728 Chr.6: 30552562-30552562 on Build GRCh38
rs762720597 Chr.6: 30552603-30552603 on Build GRCh38
rs767555737 Chr.6: 30552445-30552445 on Build GRCh38
rs763947588 Chr.6: 30552613-30552613 on Build GRCh38
rs149298420 Chr.6: 30552571-30552571 on Build GRCh38
rs774114882 Chr.6: 30552626-30552626 on Build GRCh38
rs755802323 Chr.6: 30552649-30552649 on Build GRCh38
rs73727440 Chr.6: 30552471-30552471 on Build GRCh38
rs780822937 Chr.6: 30552516-30552516 on Build GRCh38
rs145669779 Chr.6: 30552582-30552582 on Build GRCh38
rs576124674 Chr.6: 30552448-30552448 on Build GRCh38
rs755682042 Chr.6: 30552522-30552522 on Build GRCh38
rs767165987 Chr.6: 30552643-30552643 on Build GRCh38
rs775667118 Chr.6: 30552427-30552427 on Build GRCh38
rs578228176 Chr.6: 30552446-30552446 on Build GRCh38
rs780470520 Chr.6: 30552530-30552530 on Build GRCh38
rs769051259 Chr.6: 30552557-30552557 on Build GRCh38
rs146068139 Chr.6: 30552644-30552644 on Build GRCh38
rs752372831 Chr.6: 30552479-30552479 on Build GRCh38
rs745533955 Chr.6: 30552521-30552521 on Build GRCh38
rs765116090 Chr.6: 30552477-30552477 on Build GRCh38
rs761949432 Chr.6: 30552444-30552444 on Build GRCh38
rs138612204 Chr.6: 30552567-30552567 on Build GRCh38
rs771003791 Chr.6: 30552592-30552592 on Build GRCh38
rs776901989 Chr.6: 30552598-30552598 on Build GRCh38
rs200336607 Chr.6: 30552547-30552547 on Build GRCh38
rs60791005 Chr.6: 30552438-30552438 on Build GRCh38
rs762642839 Chr.6: 30552502-30552502 on Build GRCh38
rs756865539 Chr.6: 30552508-30552508 on Build GRCh38
rs758372510 Chr.6: 30552487-30552487 on Build GRCh38
rs768976260 Chr.6: 30552433-30552433 on Build GRCh38
rs370439517 Chr.6: 30552565-30552565 on Build GRCh38
rs749832407 Chr.6: 30552432-30552432 on Build GRCh38

View Full Product Details