Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 19635034-19635289 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMIP
Entrez Gene ID
51291
Gene Name
GEM interacting protein
Gene Aliases
ARHGAP46
UniGene
Hs.49427
Gene Chromosome Location
Chr.19: 19629473-19643667 on Build GRCh38
Transcripts
NM_001288998.1 NM_001288999.1 NM_016573.3 XM_005259927.2 XM_011528048.2 XM_017026861.1 XM_017026862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3101607 c.1720G>A p.E574K Chr.19: 19635054-19635054 on Build GRCh38
3970855 c.1725A>C p.I575I Chr.19: 19635049-19635049 on Build GRCh38
4131508 c.1570A>C p.T524P Chr.19: 19635204-19635204 on Build GRCh38
6195681 c.1624C>T p.L542F Chr.19: 19635150-19635150 on Build GRCh38
3959762 c.1641C>A p.P547P Chr.19: 19635133-19635133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750699499 Chr.19: 19635084-19635084 on Build GRCh38
rs1030883828 Chr.19: 19635229-19635229 on Build GRCh38
rs546665082 Chr.19: 19635091-19635091 on Build GRCh38
rs749439675 Chr.19: 19635094-19635094 on Build GRCh38
rs750870350 Chr.19: 19635232-19635232 on Build GRCh38
rs900228246 Chr.19: 19635083-19635083 on Build GRCh38
rs762442923 Chr.19: 19635055-19635055 on Build GRCh38
rs372547969 Chr.19: 19635153-19635153 on Build GRCh38
rs755087967 Chr.19: 19635100-19635100 on Build GRCh38
rs967999793 Chr.19: 19635244-19635244 on Build GRCh38
rs564904493 Chr.19: 19635242-19635242 on Build GRCh38
rs150339340 Chr.19: 19635102-19635102 on Build GRCh38
rs776160578 Chr.19: 19635204-19635217 on Build GRCh38
rs770724061 Chr.19: 19635140-19635140 on Build GRCh38
rs767951734 Chr.19: 19635069-19635069 on Build GRCh38
rs1009615135 Chr.19: 19635217-19635217 on Build GRCh38
rs762399402 Chr.19: 19635220-19635220 on Build GRCh38
rs569483813 Chr.19: 19635191-19635191 on Build GRCh38
rs761110843 Chr.19: 19635241-19635241 on Build GRCh38
rs34970769 Chr.19: 19635092-19635092 on Build GRCh38
rs752080986 Chr.19: 19635067-19635067 on Build GRCh38
rs762495802 Chr.19: 19635187-19635187 on Build GRCh38
rs527487519 Chr.19: 19635260-19635260 on Build GRCh38
rs779072696 Chr.19: 19635265-19635265 on Build GRCh38
rs1021009561 Chr.19: 19635219-19635219 on Build GRCh38
rs754083289 Chr.19: 19635254-19635254 on Build GRCh38
rs370305251 Chr.19: 19635123-19635123 on Build GRCh38
rs367726683 Chr.19: 19635155-19635155 on Build GRCh38
rs775107487 Chr.19: 19635169-19635169 on Build GRCh38
rs549666252 Chr.19: 19635108-19635108 on Build GRCh38
rs896559833 Chr.19: 19635192-19635192 on Build GRCh38
rs767848220 Chr.19: 19635221-19635221 on Build GRCh38
rs773690648 Chr.19: 19635204-19635204 on Build GRCh38
rs781052979 Chr.19: 19635141-19635141 on Build GRCh38
rs142924319 Chr.19: 19635151-19635151 on Build GRCh38
rs745781390 Chr.19: 19635058-19635058 on Build GRCh38
rs954823244 Chr.19: 19635127-19635127 on Build GRCh38
rs757599218 Chr.19: 19635068-19635068 on Build GRCh38

View Full Product Details